Merck
  • The use of 2-D gels to identify novel protein-protein interactions in the cochlea.
The use of 2-D gels to identify novel protein-protein interactions in the cochlea.
Methods in molecular biology (Clifton, N.J.) (2008-10-08)
Thandavarayan Kathiresan, Margaret C Harvey, Bernd H A Sokolowski
ABSTRACT
Functional proteomics comprises a wide range of technologies for the identification of novel protein-protein interactions and biological markers. Studies of protein-protein interactions have gained from the development of techniques and technologies such as immunoprecipitation, preparative two-dimensional (2-D) gel electrophoresis for peptide mass fingerprinting (PMF), using matrix-assisted laser desorption ionization time-of-flight (MALDI-TOF) mass spectrometry, and liquid chromatography-tandem mass spectrometry (LC-MS/MS). These applications enabled the discovery of putative protein partners without a priori knowledge of which one(s) might be relevant. Here, we report the methods by which membrane proteins are isolated from cochlear tissues and prepared for identification by mass spectrometry techniques.
Product Description
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 500 μL, pkg of 40 ea (5 bags × 8 plates)
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 1000 μL, pkg of 80 ea (10 bags × 8 plates)
Eppendorf® DNA LoBind tubes, capacity 1.5 mL, PCR clean, pkg of 250 ea (5 x 50ea)
Eppendorf® Protein LoBind Microplate 384/V-PP, PCR clean, V-bottom clear wells, white border, pack of 240 ea (10 bags × 24 plates)
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 500 μL, pkg of 40 ea (5 bags × 8 plates)
Eppendorf® Deepwell plates, DNA LoBind, 384 wells, white plate, conical bottom, colorless wells, capacity 200 μL, pack of 40 ea (5 bags × 8 plates)
Eppendorf® Deepwell plates, DNA LoBind, 384 wells, white plate, conical bottom, colorless wells, capacity 200 μL, pack of 120 ea (10 bags × 12 plates)
Eppendorf® DNA LoBind tubes, capacity 0.5 mL, PCR clean, pkg of 250 ea (5 x 50ea)
Eppendorf® DNA LoBind tubes, capacity 2.0 mL, PCR clean, pkg of 250 ea (5 x 50ea)
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 500 μL, pkg of 120 ea (10 bags × 12 plates)
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 1000 μL, pkg of 120 ea (10 bags × 12 plates)
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 1000 μL, pkg of 120 ea (10 bags × 12 plates)
Eppendorf® Deepwell plates, Protein LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 500 μL, pkg of 80 ea (10 bags × 8 plates)
Eppendorf® Deepwell plates, Protein LoBind, 96 wells, yellow plate, conical bottom, colorless wells, capacity 500 μL, pkg of 40 ea (5 bags × 8 plates)
Eppendorf® Deepwell plates, Protein LoBind, 96 wells, yellow plate, conical bottom, colorless wells, capacity 500 μL, pkg of 20 ea (5 bags × 4 plates)
Eppendorf® Protein LoBind tubes, capacity 1.5 mL, PCR clean, pkg of 100 ea (2 x 50ea)
Eppendorf® Protein LoBind tubes, capacity 0.5 mL, PCR clean, pkg of 100 ea (2 x 50ea)
Eppendorf® Protein LoBind tubes, capacity 2.0 mL, PCR clean, pkg of 100 ea (2 x 50ea)