Merck
  • Naturally-Occurring Antibodies Against Bim are Decreased in Alzheimer's Disease and Attenuate AD-type Pathology in a Mouse Model.
Naturally-Occurring Antibodies Against Bim are Decreased in Alzheimer's Disease and Attenuate AD-type Pathology in a Mouse Model.
Neuroscience bulletin (2022-05-16)
Jie-Ming Jian, Dong-Yu Fan, Ding-Yuan Tian, Yuan Cheng, Pu-Yang Sun, Cheng-Rong Tan, Gui-Hua Zeng, Chen-Yang He, Ye-Ran Wang, Jie Zhu, Xiu-Qing Yao, Yan-Jiang Wang, Yu-Hui Liu
ABSTRACT
Increased neuronal apoptosis is an important pathological feature of Alzheimer's disease (AD). The Bcl-2-interacting mediator of cell death (Bim) mediates amyloid-beta (Aβ)-induced neuronal apoptosis. Naturally-occurring antibodies against Bim (NAbs-Bim) exist in human blood, with their levels and functions unknown in AD. In this study, we found that circulating NAbs-Bim were decreased in AD patients. Plasma levels of NAbs-Bim were negatively associated with brain amyloid burden and positively associated with cognitive functions. Furthermore, NAbs-Bim purified from intravenous immunoglobulin rescued the behavioral deficits and ameliorated Aβ deposition, tau hyperphosphorylation, microgliosis, and neuronal apoptosis in APP/PS1 mice. In vitro investigations demonstrated that NAbs-Bim were neuroprotective against AD through neutralizing Bim-directed neuronal apoptosis and the amyloidogenic processing of amyloid precursor protein. These findings indicate that the decrease of NAbs-Bim might contribute to the pathogenesis of AD and immunotherapies targeting Bim hold promise for the treatment of AD.
Sigma-Aldrich
Anti-VAMP1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Neprilysin Antibody (Neutral Endopeptidase, Nep), serum, Chemicon®
Sigma-Aldrich
Anti-Synapsin-1 Antibody, clone 10.22, clone 10.22, from mouse
Sigma-Aldrich
Anti-APP A4 Antibody, a.a. 66-81 of APP {NT}, clone 22C11, clone 22C11, Chemicon®, from mouse
Sigma-Aldrich
Anti-SNAP-25 Antibody, clone SP14 (Ascites Free), clone SP14, from mouse
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-BACE 1, N-Terminus (46-62) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Tau Antibody, a.a. 210-241, clone Tau-5, ascites fluid, clone Tau-5, Chemicon®
Sigma-Aldrich
Anti-PSD95 Antibody, clone K28/43, clone K28/43, from mouse