Skip to Content
Merck
All Photos(4)

Key Documents

MAB331-C

Sigma-Aldrich

Anti-SNAP-25 Antibody, clone SP14 (Ascites Free)

clone SP14, from mouse

Synonym(s):

Synaptosomal-associated protein 25, SNAP-25, Super protein, SUP, Synaptosomal-associated 25 kDa protein, SNAP-25

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

SP14, monoclonal

species reactivity

monkey, bovine, mouse, rat, guinea pig, human

technique(s)

activity assay: suitable
electron microscopy: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... SNAP25(6616)

General description

Synaptosomal-associated protein 25 (UniProt P60880; also known as SNAP-25, SUP, Super protein, Synaptosomal-associated 25 kDa protein) is encoded by the SNAP25 (also known as SNAP) gene (Gene ID 6616) in human. SNAP-25 belongs to the superfamily of soluble N-ethylmaleimide-sensitive factor (NSF) attachment protein (SNAP) receptor (SNARE) proteins involved in mediating vesicle fusion events. Target or t-SNAREs located in the membranes of target compartments form stable subcomplexes and engage vesicle or v-SNAREs in the membranes of transport vesicles to facilitate the fusion. SNAP-25 is a t-SNARE protein of the trans-SNARE complex that executes fusion between the synaptic vesicle and plasma membrane. SANP-25 does not have a transmembrane domain and is anchored to the cytosolic face of membranes via palmitoyl side chains covalently linked to cysteine residues.

Specificity

Clone SP41 recognizes both SNAP-25 isoforms A & B. Clone SP41 is reactive toward endogenous or exogenously expressed SNAP-25 in COS cells, but not SNAP-25 expressed in E. coli, indicative of a posttranslational modification-dependent antigenicity.

Immunogen

Epitope: N-terminal half
Human brain proteins immunoprecipitated with EP10 (Cat. No. MAB332).

Application

Anti-SNAP-25 Antibody, clone SP14 (Ascites Free) is an antibody against SNAP-25 for use in Western Blotting, Immunohistochemistry (Paraffin), Electron Microscopy and Activity Assay.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blotting in human brain tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected SNAP-25 in 10 µg of human brain tissue lysate.

Target description

~25 kDa observed

Linkage

Replaces: MAB331

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Most peptide-containing sensory neurons lack proteins for exocytotic release and vesicular transport of glutamate.
Morris, JL; Konig, P; Shimizu, T; Jobling, P; Gibbins, IL
The Journal of Comparative Neurology null
Calpain activity contributes to the control of SNAP-25 levels in neurons.
Grumelli, C; Berghuis, P; Pozzi, D; Caleo, M; Antonucci, F; Bonanno, G; Carmignoto et al.
Molecular and Cellular Neurosciences null
Autoregulation in PC12 cells via P2Y receptors: Evidence for non-exocytotic nucleotide release from neuroendocrine cells.
Hussl, S; Kubista, H; Boehm, S
Purinergic Signaling null
Increased levels of SNAP-25 and synaptophysin in the dorsolateral prefrontal cortex in bipolar I disorder.
Scarr, E; Gray, L; Keriakous, D; Robinson, PJ; Dean, B
Bipolar Disorders null
Fatal familial insomnia with an unusual prion protein deposition pattern: an autopsy report with an experimental transmission study.
Sasaki, K; Doh-ura, K; Wakisaka, Y; Tomoda, H; Iwaki, T
Neuropathology and Applied Neurobiology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service