Merck
STING nuclear partners contribute to innate immune signaling responses.
iScience (2021-09-21)
Charles R Dixon, Poonam Malik, Jose I de Las Heras, Natalia Saiz-Ros, Flavia de Lima Alves, Mark Tingey, Eleanor Gaunt, A Christine Richardson, David A Kelly, Martin W Goldberg, Greg J Towers, Weidong Yang, Juri Rappsilber, Paul Digard, Eric C Schirmer
ABSTRACT
STimulator of INterferon Genes (STING) is an adaptor for cytoplasmic DNA sensing by cGAMP/cGAS that helps trigger innate immune responses (IIRs). Although STING is mostly localized in the ER, we find a separate inner nuclear membrane pool of STING that increases mobility and redistributes to the outer nuclear membrane upon IIR stimulation by transfected dsDNA or dsRNA mimic poly(I:C). Immunoprecipitation of STING from isolated nuclear envelopes coupled with mass spectrometry revealed a distinct nuclear envelope-STING proteome consisting of known nuclear membrane proteins and enriched in DNA- and RNA-binding proteins. Seventeen of these nuclear envelope STING partners are known to bind direct interactors of IRF3/7 transcription factors, and testing a subset of these revealed STING partners SYNCRIP, MEN1, DDX5, snRNP70, RPS27a, and AATF as novel modulators of dsDNA-triggered IIRs. Moreover, we find that SYNCRIP is a novel antagonist of the RNA virus, influenza A, potentially shedding light on reports of STING inhibition of RNA viruses.
Sigma-Aldrich
Trypsin Singles, Proteomics Grade
Sigma-Aldrich
Leupeptin, synthetic, ≥85% (HPLC)
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Anti-MB21D1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-hnRNP Q Antibody, clone 7A11.2, clone 7A11.2, from mouse
Sigma-Aldrich
Polyinosinic–polycytidylic acid sodium salt, TLR ligand tested
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Toluidine Blue, 8.74% (ZN (THEORY)), for microscopy (Hist., Vit.)
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)