Skip to Content
Merck
All Photos(1)

Key Documents

T7575

Sigma-Aldrich

Trypsin Singles, Proteomics Grade

Synonym(s):

Proteomics grade, Trypsin, TPCK treated

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.56
Pricing and availability is not currently available.

grade

Proteomics Grade

form

ready-to-use solution

specific activity

≥10,000 BAEE units/mg protein

mol wt

average mol wt 23.29 kDa

optimum pH

~8.0

storage temp.

2-8°C

General description

Experience all of the advantages of Proteomics Grade Trypsin in a convenient, single-use 1 μg package.

Trypsin Singles, Proteomics Grade exhibit excellent proteolytic efficiency, generating more tryptic peptides leading to greater sequence coverage of your peptide of interest. Mass spectra are significantly simplified due to the reduced number of interfering autolytic peaks and their ambiguous adducts. Proteomics Grade Trypsin has been extensively purified from porcine pancreas to enable accurate and precise cleavage on the carboxylic side of Arg and Lys residues. The enzyme has been exhaustively processed by reductive methylation to minimize autolysis and chymotryptic activity has been quenched by TPCK treatment.[1] Further purification steps including affinity chromatography and drying from dilute acid produce a highly purified, high specific activity trypsin purposely suited for the demanding criteria of proteomics research, and designed to function in either solution or gel-based digests.

Application

Trypsin Singles, Proteomics Grade have been used for protein digestion to analyse generated peptides via MALDI-TOF MS (matrix assisted laser desorption ionization-time of flight mass spectrometry).[2][3] It has been used for the digestion of proteins to analyse generated peptides via LC-MS (liquid chromatography–mass spectrometry)/MS.[4][5]

Features and Benefits

  • Convenient, single-use 1 μg size
  • Eliminates repetitive pipetting
  • Reductively methylated to minimize autolytic activity
  • TPCK treated to quench chymotryptic activity
  • Extensively purified

Kit Components Also Available Separately

Product No.
Description
SDS

  • Trypsin Solubilization Reagent

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Costanzo et al.
Science (New York, N.Y.), 327(5964), 425-431 (2010-01-23)
A genome-scale genetic interaction map was constructed by examining 5.4 million gene-gene pairs for synthetic genetic interactions, generating quantitative genetic interaction profiles for approximately 75% of all genes in the budding yeast, Saccharomyces cerevisiae. A network based on genetic interaction
Zahraa I Khamis et al.
Journal of Cancer, 1, 70-79 (2010-09-16)
In situ neoplastic prostate cells are not lethal unless they become invasive and metastatic. For cells to become invasive, the prostate gland must undergo degradation of the basement membrane and disruption of the basal cell layer underneath the luminal epithelia.
Tiziana Cabras et al.
Molecular & cellular proteomics : MCP, 9(10), 2099-2108 (2010-06-30)
The acidic soluble fraction of whole saliva of type 1 diabetic children was analyzed by reversed phase (RP)(1)-HPLC-ESI-MS and compared with that of sex- and age-matched control subjects. Salivary acidic proline-rich phosphoproteins (aPRP), histatins, α-defensins, salivary cystatins, statherin, proline-rich peptide
Identification of protein binders in artworks by MALDI-TOF/TOF tandem mass spectrometry.
Tripkovic T, et al.
Talanta, 113, 49-61 (2013)
Melt With This Kiss: Paralyzing and Liquefying Venom of The Assassin Bug Pristhesancus plagipennis (Hemiptera: Reduviidae).
Walker AA, et al.
Molecular and Cellular Proteomics, 16, 552-552 (2017)

Questions

  1. How many 1 µg samples are included in the proteomics grade Trypsin kit (T7575-1KT)?

    1 answer
    1. The proteomics grade Trypsin kit (T7575-1KT) includes 96 individual 1 µg samples, each provided in a single vial.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service