Merck
  • Megabodies expand the nanobody toolkit for protein structure determination by single-particle cryo-EM.
Megabodies expand the nanobody toolkit for protein structure determination by single-particle cryo-EM.
Nature methods (2021-01-08)
Tomasz Uchański, Simonas Masiulis, Baptiste Fischer, Valentina Kalichuk, Uriel López-Sánchez, Eleftherios Zarkadas, Miriam Weckener, Andrija Sente, Philip Ward, Alexandre Wohlkönig, Thomas Zögg, Han Remaut, James H Naismith, Hugues Nury, Wim Vranken, A Radu Aricescu, Els Pardon, Jan Steyaert
ABSTRACT
Nanobodies are popular and versatile tools for structural biology. They have a compact single immunoglobulin domain organization, bind target proteins with high affinities while reducing their conformational heterogeneity and stabilize multi-protein complexes. Here we demonstrate that engineered nanobodies can also help overcome two major obstacles that limit the resolution of single-particle cryo-electron microscopy reconstructions: particle size and preferential orientation at the water-air interfaces. We have developed and characterized constructs, termed megabodies, by grafting nanobodies onto selected protein scaffolds to increase their molecular weight while retaining the full antigen-binding specificity and affinity. We show that the megabody design principles are applicable to different scaffold proteins and recognition domains of compatible geometries and are amenable for efficient selection from yeast display libraries. Moreover, we demonstrate that megabodies can be used to obtain three-dimensional reconstructions for membrane proteins that suffer from severe preferential orientation or are otherwise too small to allow accurate particle alignment.
Sigma-Aldrich
2,2-Dimethyl-1,3-propanediol, 99%
Sigma-Aldrich
Nonaethylene glycol monododecyl ether, nonionic surfactant
Sigma-Aldrich
Membrane Scaffold Protein 1E3D1, recombinant, expressed in E. coli
Sigma-Aldrich
Membrane Scaffold Protein 1D1, recombinant, expressed in E. coli
Sigma-Aldrich
Membrane Scaffold Protein 2N2, recombinant, expressed in E. coli, MSP1D1-MSP1D2 fusion protein
Sigma-Aldrich
Lysozyme from chicken egg white, powder or granules, ≥90 %, ≥39,000 units/mg protein
Sigma-Aldrich
Maltose solution, for molecular biology, BioReagent, ~20% in H2O