Merck
  • Phototocatalytic lithography of poly(propylene sulfide) block copolymers: toward high-throughput nanolithography for biomolecular arraying applications.
Phototocatalytic lithography of poly(propylene sulfide) block copolymers: toward high-throughput nanolithography for biomolecular arraying applications.
Langmuir : the ACS journal of surfaces and colloids (2008-12-31)
Jane P Bearinger, Gary Stone, Amy L Hiddessen, Lawrence C Dugan, Ligang Wu, Philip Hailey, James W Conway, Tobias Kuenzler, Lydia Feller, Simona Cerritelli, Jeffrey A Hubbell
ABSTRACT
Photocatalytic lithography (PCL) is an inexpensive, fast, and robust method of oxidizing surface chemical moieties to produce patterned substrates. This technique has utility in basic biological research as well as various biochip applications. We report on porphyrin-based PCL for patterning poly(propylene sulfide) block copolymer films on gold substrates on the micrometer and submicrometer scales. We confirm chemical patterning with imaging ToF-SIMS and low-voltage SEM. Biomolecular patterning on micrometer and submicrometer scales is demonstrated with proteins, protein-linked beads. and fluorescently labeled proteins.
Sigma-Aldrich
Magnesium phthalocyanine, Dye content 90 %
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, denatured, reagent grade, denatured
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Ethanol, 94.8-95.8%
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Reagent Alcohol, denatured, suitable for HPLC
Sigma-Aldrich
Reagent Alcohol, reagent grade
Sigma-Aldrich
Chlorophyllin sodium copper salt