Merck
  • Syndecan-4 tunes cell mechanics by activating the kindlin-integrin-RhoA pathway.
Syndecan-4 tunes cell mechanics by activating the kindlin-integrin-RhoA pathway.
Nature materials (2020-01-08)
Antonios Chronopoulos, Stephen D Thorpe, Ernesto Cortes, Dariusz Lachowski, Alistair J Rice, Vasyl V Mykuliak, Tomasz Róg, David A Lee, Vesa P Hytönen, Armando E Del Río Hernández
ABSTRACT
Extensive research over the past decades has identified integrins to be the primary transmembrane receptors that enable cells to respond to external mechanical cues. We reveal here a mechanism whereby syndecan-4 tunes cell mechanics in response to localized tension via a coordinated mechanochemical signalling response that involves activation of two other receptors: epidermal growth factor receptor and β1 integrin. Tension on syndecan-4 induces cell-wide activation of the kindlin-2/β1 integrin/RhoA axis in a PI3K-dependent manner. Furthermore, syndecan-4-mediated tension at the cell-extracellular matrix interface is required for yes-associated protein activation. Extracellular tension on syndecan-4 triggers a conformational change in the cytoplasmic domain, the variable region of which is indispensable for the mechanical adaptation to force, facilitating the assembly of a syndecan-4/α-actinin/F-actin molecular scaffold at the bead adhesion. This mechanotransduction pathway for syndecan-4 should have immediate implications for the broader field of mechanobiology.
Sigma-Aldrich
Monoclonal Anti-α-Actinin antibody produced in mouse, clone BM-75.2, ascites fluid
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
1,2-Dilinoleoyl-3-palmitoyl-rac-glycerol, ≥95% (TLC), liquid
Sigma-Aldrich
Latrunculin A, Latrunculia magnifica, InSolution, ≥85%, 1 mM in DMSO, marine toxin that disrupts microfilament organization in cultured cells
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine, 15 mM HEPES, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Triton X-100, for molecular biology
Sigma-Aldrich
Latrunculin A, from sea sponge, ≥85% (HPLC), waxy solid
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
LY-294,002 hydrochloride, solid, ≥98% (HPLC)
Sigma-Aldrich
Anti-Integrin β1 Antibody, activated, clone HUTS-4, Azide Free, clone HUTS-4, Chemicon®, from mouse
Sigma-Aldrich
Anti-Fibronectin Frag Antibody, cell attachment fragment, clone 3E3, clone 3E3, Chemicon®, from mouse
Sigma-Aldrich
Mouse IgG1-k Negative Control, clone MOPC-21, Azide Free Antibody, clone MOPC-21, from mouse
Sigma-Aldrich
Y-27632, InSolution, ≥95%, reversible, inhibitor of Rho kinases
Sigma-Aldrich
Anti-Kindlin-2 Antibody, clone 3A3, clone 3A3, from mouse