Skip to Content
Merck
All Photos(1)

Key Documents

MABF1081Z

Sigma-Aldrich

Mouse IgG1-k Negative Control, clone MOPC-21, Azide Free Antibody

clone MOPC-21, from mouse

Synonym(s):

IgG1-k

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.42

biological source

mouse

Quality Level

antibody form

purified antibody

clone

MOPC-21, monoclonal

technique(s)

flow cytometry: suitable

isotype

IgG1

target post-translational modification

unmodified

General description

The MOPC-21 immunoglobulin is useful as an isotype-matched control. The MOPC-21 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG1 antibodies.

Immunogen

Immunogen information for this antibody is proprietary information.

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This mouse monoclonal Mouse IgG1-k Negative Control, clone MOPC-21, Azide Free, Cat. No. MABF1081Z is validated for use in Flow Cytometry.

Physical form

Affinity Chromatography
Format: Purified
Purified Mouse monoclonal containing 10 mM NaH2PO4, 150 mM NaCl, pH7.2. This monoclonal antibody was purified from tissue culture supernatant via affinity chromatography.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Note: It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eveliina Pollari et al.
Journal of neuroinflammation, 8, 74-74 (2011-06-30)
Granulocyte colony stimulating factor (GCSF) is protective in animal models of various neurodegenerative diseases. We investigated whether pegfilgrastim, GCSF with sustained action, is protective in a mouse model of amyotrophic lateral sclerosis (ALS). ALS is a fatal neurodegenerative disease with
Kinola J N Williams et al.
Microbiology and immunology, 60(11), 754-769 (2016-11-20)
In this study, the effect of active infection with vaccinia virus Western Reserve (VACV WR) on expression of C-type lectin domain family 2 (CLEC2D), a ligand of the human NK cell inhibitory receptor NKR-P1, was examined. As predicted, VACV infection
Claire E Johnston et al.
PloS one, 7(10), e48195-e48195 (2012-11-08)
Chicken eggs are the main source of human Salmonella enterica serovar Enteritidis infection. S. Enteritidis infects the oviduct and ovary of the chicken leading to infection of developing eggs. Therefore, control in poultry production is a major public health priority.
Li Li et al.
Blood, 104(3), 815-821 (2004-04-20)
The lymphoid follicle is a specialized microenvironment for the differentiation of antigen (Ag)-activated B cells; the major stromal cell components in lymphoid follicle are the follicular dendritic cells (FDCs). At the same time, most of the B-cell lymphomas originate from
Osama M A Ibrahim et al.
Investigative ophthalmology & visual science, 53(4), 2403-2413 (2012-03-14)
To investigate the lipid and DNA oxidative stress as well as corneal and retinal effects after ultraviolet B (UV-B) exposure in mice, with or without silicon hydrogel soft contact lenses (SCL). Twenty-eight C57BL6-strain male mice were divided into four groups:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service