Skip to Content
Merck
All Photos(4)

Key Documents

MAB2617

Sigma-Aldrich

Anti-Kindlin-2 Antibody, clone 3A3

clone 3A3, from mouse

Synonym(s):

Mitogen-inducible gene 2 protein, Pleckstrin homology domain-containing family C member 1, fermitin family homolog 2, kindlin 2, mitogen inducible gene 2 protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3A3, monoclonal

species reactivity

human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), rat (98% sequence homology), mouse (98% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FERMT2(10979)

General description

The Kindlin family is a group of focal adhesion proteins that have become a point of interest because of their ability to bind and activate integrins. Abnormalities in this ability have linked Kindlin to several diseases, both inherited and acquired, such as Kindler syndrome, cancer, and leukocyte adhesion deficiency. The isoform Kindlin-2 specifically participates in the recruitment of migfilin protein to focal adhesion sites for cell-ECM interactions, coordinates the manufacture of actin and modifications to cell shape. It is also actively involved in connecting ECM adhesion sites and the actin cytoskeleton (Shi, 2007). Kindlin-2 is pervasively expressed throughout tissue.

Specificity

The antibody recognizes Kindlin-2.

Immunogen

Epitope: Unknown
GST-tagged recombinant protein corresponding to human Kindlin-2.

Application

Research Category
Cell Structure
Research Sub Category
Cytoskeleton
This Anti-Kindlin-2 Antibody, clone 3A3 is validated for use in WB, IC for the detection of Kindlin-2.

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 0.5 µg/ml of this antibody detected Kindlin-2 in 10 µg of HeLa cell lysate.

Target description

~ 70 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Lin et al.
Journal of experimental & clinical cancer research : CR, 36(1), 134-134 (2017-10-04)
Kindlin-2 is a member of the focal adhesion protein family that regulates invasion and metastasis in multiple malignancies; however, little is known about the role of Kindlin-2 in hepatocellular carcinoma (HCC) progression. Immunohistochemistry was used to investigate Kindlin-2 expression in
Huanqing Gao et al.
JCI insight, 4(13) (2019-07-12)
Kindlin-2 regulates integrin-mediated cell adhesion to and migration on the extracellular matrix. Our recent studies demonstrate important roles of kindlin-2 in regulation of mesenchymal stem cell differentiation and skeletal development. In this study, we generated adipose tissue-specific conditional knockout of
Lihua Qi et al.
Cell death & disease, 10(12), 890-890 (2019-11-27)
Kindlin-2 plays an important role in the regulation of cardiac structure and function. Depletion of Kindlin-2 contributes to cardiac hypertrophy and progressive heart failure, however, the precise mechanisms involved in this process remain unclear. GATA4 is a critical transcription factor
Lihua Qi et al.
FEBS letters, 589(16), 2155-2162 (2015-07-06)
Kindlin-2, as an integrin-interacting protein, was known to be required for the maintenance of cardiac structure and function in zebrafish. However, the mechanism remains unclear. We found that Kindlin-2 interacts and colocalizes with α-actinin-2 at the Z-disc of mouse cardiac
Khalid Sossey-Alaoui et al.
Cancer research, 77(18), 5129-5141 (2017-07-09)
Interplay between tumor cells and host cells in the tumor microenvironment dictates the development of all cancers. In breast cancer, malignant cells educate host macrophages to adopt a protumorigenic phenotype. In this study, we show how the integrin-regulatory protein kindlin-2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service