Merck
  • Role of the Ang2-Tie2 Axis in Vascular Damage Driven by High Glucose or Nucleoside Diphosphate Kinase B Deficiency.
Role of the Ang2-Tie2 Axis in Vascular Damage Driven by High Glucose or Nucleoside Diphosphate Kinase B Deficiency.
International journal of molecular sciences (2020-05-30)
Anupriya Chatterjee, Rachana Eshwaran, Hongpeng Huang, Di Zhao, Martina Schmidt, Thomas Wieland, Yuxi Feng
ABSTRACT
Ablation of nucleoside diphosphate kinase B (NDPK-B) in mice causes a breakdown of the neurovascular unit in the retina, mimicking diabetic retinopathy. The NDPK-B deficiency-induced vascular damage is mediated by excessive angiopoietin 2 (Ang2). Herein, the potential involvement of its receptor, Tie2, was investigated. NDPK-B-deficient mouse retinas showed an upregulation of Tie2, specifically in the deep capillary layer. A similar upregulation of Tie2 was observed in cultured endothelial cells (ECs) from different origins upon NDPK-B depletion, whereas high glucose (HG) treatment did not alter Tie2 expression. Immunofluorescence staining and subcellular fractionation showed that the majority of Tie2 upregulation occurred at the plasma membrane. Similar to HG, however, NDPK-B depletion reduced Tie2 tyrosine phosphorylation. Compared to HG, a stronger increase of Ang2 was observed in NDPK-B depleted ECs. Treatment of ECs with soluble Tie2 or siRNA-mediated Tie2 knockdown attenuated NDPK-B depletion- but not HG-induced Ang2 upregulation. Like NDPK-B depletion, overexpression of recombinant Ang2 in ECs enhanced Ang2 secretion and concomitantly promoted the upregulation of Tie2. Thus, we identified a new mechanism showing that after reaching a threshold level of secretion, Ang2 sustains its own expression and secretion by a Tie2-dependent positive feedback loop.
Sigma-Aldrich
Anti-Goat IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
Anti-Tie2/TEK Antibody, clone Ab33, clone Ab33, Upstate®, from mouse
Millipore
Gelatin from porcine skin, suitable for microbiology, low gel strength