Skip to Content
Merck
All Photos(1)

Documents

48720

Millipore

Gelatin from porcine skin

suitable for microbiology, low gel strength

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41106212
NACRES:
NA.85

biological source

Porcine

Quality Level

sterility

non-sterile

form

powder

quality

low gel strength

ign. residue

≤2%

loss

≤15% loss on drying

pH

4.0-6.0 (50 °C, 67 mg/mL in H2O)

gel strength

90-110 g Bloom (67 mg/ml water)

solubility

H2O: 67 mg/mL at 50 °C, slightly hazy, light brownish-yellow

anion traces

chloride (Cl-): ≤2000 mg/kg

cation traces

Ca: ≤2000 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤10 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤500 mg/kg
Mg: ≤500 mg/kg
Mn: ≤5 mg/kg
Na: ≤3000 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤10 mg/kg

application(s)

microbiology

Looking for similar products? Visit Product Comparison Guide

Application

This product is recommended for use as a cell culture substratum at 1-5 μg/cm2 or 0.5-50 μg/mL. The optimal concentration does depend on cell type as well as the application and research objectives.

Gelatin has been used in many applications. It has use in coating cell culture to improve attachment of cells, being added to PCR to stabilize Taq DNA, as a blocking reagent in Western blotting, ELISA, and immunochemistry, and as a component of media for species differentiation in bacteriology. As a biocompatible polymer, it has used as a delivery vehicle for release of active biomolecules and in generation of scaffolds for tissue engineering applications. In the pharmaceutical industry, geltan can be used as a suspending and encapsulating agent, among other applications.

Components

Gelatin is a heterogeneous mixture of water-soluble proteins of high average molecular masses, present in collagen. Proteins are extracted by boiling the relevant skin, tendons, ligaments, bones, etc. in water. Type A gelatin is derived from acid-cured tissue. Type B is derived from lime-cured tissue.

Caution

Dry gelatin, when stored in airtight containers at room temperature, will remain unchanged for many years. When heated at 100°C in the presence of air, it swells becomes soft and disintegrates to a carbonaceous mass with evolution of pyridine bases and ammonia.

Other Notes

This product is derived from porcine skin. Gelatin is soluble in hot than in cold water. It is practically insoluble in most organic solvents such as alcohol, chloroform, carbon disulfide, carbon tetrachloride, ether, benzene, acetone, and oils. The Bloom number, determined by the Bloom gelometer, is an indication of the strength of a gel formed from a solution of the known concentration. The Bloom number is proportional to the average molecular mass. Bloom numbers of porcine skin Gelatin vary from 90 to 300 g. This product has a gel strength of 60. Manufactured by Gelita AG

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anupriya Chatterjee et al.
International journal of molecular sciences, 21(10) (2020-05-30)
Ablation of nucleoside diphosphate kinase B (NDPK-B) in mice causes a breakdown of the neurovascular unit in the retina, mimicking diabetic retinopathy. The NDPK-B deficiency-induced vascular damage is mediated by excessive angiopoietin 2 (Ang2). Herein, the potential involvement of its
Anupriya Chatterjee et al.
Cells, 9(10) (2020-10-23)
Our previous studies identified that retinal endothelial damage caused by hyperglycemia or nucleoside diphosphate kinase-B (NDPK-B) deficiency is linked to elevation of angiopoietin-2 (Ang-2) and the activation of the hexosamine biosynthesis pathway (HBP). Herein, we investigated how NDPK-B is involved
Leslie Crews et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 28(16), 4250-4260 (2008-04-18)
Altered expression and mutations in alpha-synuclein (alpha-syn) have been linked to Parkinson's disease (PD) and related disorders. The neurological alterations in PD patients have been associated with degeneration of dopaminergic cells and other neuronal populations. Moreover, recent studies in murine
Monica A Serban et al.
Methods in molecular biology (Clifton, N.J.), 1001, 133-143 (2013-03-16)
Tissue engineering involves the concerted action of biomaterials, cells, and growth factors. Kidney -regeneration relies on the same combination of ingredients. Here, we describe an example of gelatin-based biomaterial preparation and its evaluation in the context of kidney biocompatibility and
Takeo Furuya et al.
The journal of spinal cord medicine, 36(2), 134-139 (2013-07-03)
Besides stimulating angiogenesis or cell survival, basic fibroblast growth factor (bFGF) has the potential for protecting neurons in the injured spinal cord. To investigate the effects of a sustained-release system of bFGF from gelatin hydrogel (GH) in a rat spinal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service