Skip to Content
Merck
All Photos(1)

Key Documents

131695

Sigma-Aldrich

Pyrimidine

≥98.0%

Synonym(s):

1,3-Diazine, Metadiazine, m-Diazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4N2
CAS Number:
Molecular Weight:
80.09
Beilstein:
103894
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39161701
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥98.0%

refractive index

n20/D 1.504 (lit.)

bp

123-124 °C (lit.)

mp

19-22 °C (lit.)

solubility

H2O: soluble
alcohols: soluble
diethyl ether: soluble

density

1.016 g/mL at 25 °C (lit.)

SMILES string

c1cncnc1

InChI

1S/C4H4N2/c1-2-5-4-6-3-1/h1-4H

InChI key

CZPWVGJYEJSRLH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyrimidine is a heterocyclic compound with excellent stability. It is used in manufacture of pharmaceuticals, agrochemicals and dyes. [1]

Application

Pyrimidine was used to assess the extent of pyrimidine/purine asymmetry quantitatively[2]. It was also used to study the photoinduced ion chemistry of the halogenated pyrimidines, a class of prototype radiosensitizing molecules[3].

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Vladimir Pabon-Martinez et al.
Scientific reports, 7(1), 11043-11043 (2017-09-10)
The anti-gene strategy is based on sequence-specific recognition of double-strand DNA by triplex forming (TFOs) or DNA strand invading oligonucleotides to modulate gene expression. To be efficient, the oligonucleotides (ONs) should target DNA selectively, with high affinity. Here we combined
Pyrimidine: a review on anticancer activity with key emphasis on SAR
Aastha M et al.
Future Journal of Pharmaceutical Sciences, 7, 123-123 (2021)
Elke J A Hilgers et al.
Frontiers in plant science, 9, 1331-1331 (2018-10-20)
The xylulose 5-phosphate/phosphate translocator (XPT) represents the fourth functional member of the phosphate translocator (PT) family residing in the plastid inner envelope membrane. In contrast to the other three members, little is known on the physiological role of the XPT.
Mattea Carmen Castrovilli et al.
Journal of the American Society for Mass Spectrometry, 25(3), 351-367 (2014-01-05)
In the present work, we studied the photoinduced ion chemistry of the halogenated pyrimidines, a class of prototype radiosensitizing molecules, in the energy region 9-15 eV. The work was stimulated by previous studies on inner shell site-selective fragmentation of the
R Piechocki et al.
TAG. Theoretical and applied genetics. Theoretische und angewandte Genetik, 49(6), 265-271 (1977-11-01)
The correlation between the evolutionary rates of proteins and frequencies of DNA-bases in the first and in the second position of corresponding codons was investigated.Adenine in the first and guanine in the second position showed the strongest positive correlation with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service