Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lead(II) acetate trihydrate
Synonym(s):
Lead(II) acetate trihydrate
Linear Formula:
Pb(CH
3
CO
2
)
2
· 3H
2
O
CAS Number:
6080-56-4
Molecular Weight:
379.33
EC Number:
206-104-4
Products
Product Category
salts (4)
Brand
Sigma-Aldrich (7)
Supelco (2)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Special Grade
ACS reagent (1)
Puriss P.A. (1)
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
215902
Lead(II) acetate trihydrate,
ACS reagent, ≥99%
Expand
View Pricing
32307
Lead(II) acetate trihydrate,
puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-102.0%
Expand
View Pricing
316512
Lead(II) acetate trihydrate,
99.999% trace metals basis
Expand
View Pricing
1.07375
Lead(II) acetate trihydrate,
for analysis EMSURE
®
ACS,Reag. Ph Eur
Expand
View Pricing
467863
Lead(II) acetate trihydrate,
≥99.99% trace metals basis
Expand
View Pricing
1.07372
Lead(II) acetate trihydrate,
extra pure, Technipur
®
Expand
View Pricing
915440
Lead(II) acetate trihydrate,
99.999% trace metals basis, organic soluble for perovskites
Expand
View Pricing
1.93679
Lead(II) acetate trihydrate,
EMPLURA
®
Expand
View Pricing
18-0500
Lead(II) acetate trihydrate,
JIS special grade, ≥99.5%
Expand
View Pricing