Skip to Content
Merck
All Photos(1)

Key Documents

915440

Sigma-Aldrich

Lead(II) acetate trihydrate

99.999% trace metals basis, organic soluble for perovskites

Synonym(s):

Lead diacetate trihydrate, Plumbous acetate trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb(CH3CO2)2 · 3H2O
CAS Number:
Molecular Weight:
379.33
Beilstein:
3730298
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23
Assay:
99.999% trace metals basis
form:
solid
Pricing and availability is not currently available.

Quality Level

Assay

99.999% trace metals basis

form

solid

mp

75 °C (dec.) (lit.)

SMILES string

O.O.O.CC(=O)O[PbH2]OC(C)=O

InChI

1S/2C2H4O2.3H2O.Pb/c2*1-2(3)4;;;;/h2*1H3,(H,3,4);3*1H2;/q;;;;;+2/p-2

InChI key

MCEUZMYFCCOOQO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Lead(II) Acetate Trihydrate finds application in synthesis of perovksites based photovoltaic materials[1][2]. Our perovskite grade Lead(II) Acetate Trihydrate can readily be dissolved in DMF to yield 1M solution.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1A - STOT RE 1

Target Organs

Central nervous system,Blood,Immune system,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lead acetate precursor based p-i-n perovskite solar cells with enhanced reproducibility and low hysteresis
Forgacs D, et al.
Journal of Material Chemistry A, 3, 14121-14125 (2015)
Fabricating High-Efficient Blade-Coated Perovskite Solar Cells under Ambient Condition Using Lead Acetate Trihydrate
Kong W, et al.
Solar RRL, 2(3), 1700214-1700214 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service