Skip to Content
Merck
All Photos(1)

Key Documents

1622000

USP

Stearyl alcohol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1-Octadecanol, Octadecyl alcohol, Stearyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17OH
CAS Number:
Molecular Weight:
270.49
Beilstein:
1362907
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor density

9.3 (vs air)

vapor pressure

<0.01 mmHg ( 38 °C)

API family

stearyl alcohol

autoignition temp.

842 °F

expl. lim.

~8 %

manufacturer/tradename

USP

mp

56-59 °C (lit.)

density

0.812 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCCCCCCCCCCCCCCCO

InChI

1S/C18H38O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h19H,2-18H2,1H3

InChI key

GLDOVTGHNKAZLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Stearyl alcohol USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Cetostearyl Alcohol
  • Sodium Stearyl Fumarate
  • Hydrogenated Lanolin
  • Cetyl Alcohol
  • Myristyl Alcohol
  • Oleyl Alcohol

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

383.0 °F - closed cup

Flash Point(C)

195 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cetyl Alcohol
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 46(3), 5705-5705 (2020)
Stearyl Alcohol
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 46(3), 6072-6072 (2020)
Hydrogenated Lanolin
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 39(5), 5850-5850 (2015)
Cetostearyl Alcohol
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 48(4), 5702-5702 (2015)
A Carpio et al.
Journal of chromatography. A, 1370, 25-32 (2014-12-03)
A periodic mesoporous organosilica, in particular, a phenylene-bridged material (Ph-PMO), was evaluated for the first time as a sorbent for retaining and eluting fenuron, simazine, atrazine, carbaryl and terbutryn in grape must by solid phase extraction (SPE) prior to their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service