Skip to Content
Merck
All Photos(1)

Documents

S1350000

Stearyl alcohol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-Octadecanol, Octadecyl alcohol, Stearyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17OH
CAS Number:
Molecular Weight:
270.49
Beilstein:
1362907
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor density

9.3 (vs air)

vapor pressure

<0.01 mmHg ( 38 °C)

API family

stearyl alcohol

autoignition temp.

842 °F

expl. lim.

~8 %

manufacturer/tradename

EDQM

mp

56-59 °C (lit.)

density

0.812 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCCO

InChI

1S/C18H38O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h19H,2-18H2,1H3

InChI key

GLDOVTGHNKAZLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Stearyl alcohol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

383.0 °F - closed cup

Flash Point(C)

195 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Sylvie Roke et al.
Proceedings of the National Academy of Sciences of the United States of America, 103(36), 13310-13314 (2006-08-30)
We investigate the phase behavior of surface-functionalized silica colloids at both the molecular and macroscopic levels. This investigation allows us to relate collective properties such as aggregation, gelation, and aging directly to molecular interfacial behavior. By using surface-specific vibrational spectroscopy
A Antonelli et al.
Nanotechnology, 21(42), 425101-425101 (2010-09-23)
Single-walled carbon nanotubes (SWNTs) due to their unique structural and physicochemical properties, have been proposed as delivery systems for a variety of diagnostic and therapeutic agents. However, SWNTs have proven difficult to solubilize in aqueous solution, limiting their use in
Laura Carreto et al.
Biophysical journal, 82(1 Pt 1), 530-540 (2001-12-26)
As a model for the epicuticular wax layer of plant cuticular membranes, we have studied the phase behavior of 1-tetradecanol and 1-octadecanol and their binary mixtures between 5 and 70 degrees C, using differential scanning calorimetry and Fourier transform infrared
Allergic contact dermatitis from stearyl alcohol in Efudix cream.
P D Yesudian et al.
Contact dermatitis, 45(5), 313-314 (2001-11-28)
Jörg Bauer et al.
Journal of the American Chemical Society, 127(20), 7296-7297 (2005-05-19)
Hydrophobically assisted switching phase (HASP) synthesis is a concept that allows the choice between the advantages of solid-supported chemistry and those of solution-phase synthesis. Starting from the examination of adsorption and desorption properties of hydrophobic molecules to and from reversed-phase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service