Skip to Content
Merck
All Photos(1)

Key Documents

1615956

USP

Sorbic acid

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

α-trans-γ-trans-Sorbic acid, (E,E)-1,3-Pentadiene-1-carboxylic acid, 2,4-Hexadienoic acid, 2E,4E-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, C6:2n-2,4, NSC 35405, NSC 49103, NSC 50268

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCH=CHCOOH
CAS Number:
Molecular Weight:
112.13
Beilstein:
1098547
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor pressure

0.01 mmHg ( 20 °C)

API family

sorbic acid

manufacturer/tradename

USP

mp

132-135 °C (lit.)

density

1.2 g/cm3 at 20 °C

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

C\C=C\C=C\C(O)=O

InChI

1S/C6H8O2/c1-2-3-4-5-6(7)8/h2-5H,1H3,(H,7,8)/b3-2+,5-4+

InChI key

WSWCOQWTEOXDQX-MQQKCMAXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sorbic acid USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Cranberry Fruit Juice
  • Cranberry Fruit Juice Concentrate
  • Sorbic Acid

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

G D Khandelwal et al.
Food additives and contaminants, 7(5), 685-694 (1990-09-01)
The conjugated dienoic acid structure of sorbic acid renders it susceptible to nucleophilic attack. Nucleophiles known to react with sorbic acid include sulphite ion and amines. These attack the molecule in position 5 and, in the cse of amines, cyclization
Malcolm Stratford et al.
International journal of food microbiology, 181, 40-47 (2014-05-13)
The food spoilage yeast Zygosaccharomyces bailii shows great resistance to weak-acid preservatives, including sorbic acid (2, 4-hexadienoic acid). That extreme resistance was shown to be due to population heterogeneity, with a small sub-population of cells resistant to a variety of
B L Wedzicha et al.
Food additives and contaminants, 7(5), 695-709 (1990-09-01)
In common with many other carboxylic acids, sorbic acid shows significant solubility in aqueous and non-aqueous solvents. The presence of a non-aqueous phase (e.g. fat) can markedly affect the concentration of the preservative in the aqueous phase. Solute distribution between
The genotoxicity status of sorbic acid, potassium sorbate and sodium sorbate.
F E Würgler et al.
Mutation research, 283(2), 107-111 (1992-10-01)
R Walker
Food additives and contaminants, 7(5), 671-676 (1990-09-01)
Sorbic acid and its salts have been subjected to an extensive battery of tests, including acute, short-term and chronic toxicity/carcinogenicity tests, two-generation reproduction and teratogenicity studies. These studies show that sorbic acid and sorbates have a very low level of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service