Skip to Content
Merck
All Photos(1)

Key Documents

1612700

USP

Simvastatin

United States Pharmacopeia (USP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H38O5
CAS Number:
Molecular Weight:
418.57
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

simvastatin

manufacturer/tradename

USP

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

127-132 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@]12[C@H](C[C@@H](C)C=C1C=C[C@H](C)[C@@H]2CC[C@@H]3C[C@@H](O)CC(=O)O3)OC(=O)C(C)(C)CC

InChI

1S/C25H38O5/c1-6-25(4,5)24(28)30-21-12-15(2)11-17-8-7-16(3)20(23(17)21)10-9-19-13-18(26)14-22(27)29-19/h7-8,11,15-16,18-21,23,26H,6,9-10,12-14H2,1-5H3/t15-,16-,18+,19+,20-,21-,23-/m0/s1

InChI key

RYMZZMVNJRMUDD-HGQWONQESA-N

Gene Information

human ... HMGCR(3156)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Simvastatin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Simvastatin Tablets

Biochem/physiol Actions

Simvastatin is a specific inhibitor of HMG-CoA reductase and cholesterol lowering drug.
Simvastatin is a specific inhibitor of HMG-CoA reductase, the enzyme that catalyzes the conversion of HMG-CoA to mevalonate, an early step in cholesterol biosynthesis. It is used in the treatment of hypercholesterolemia, as it reduces levels of low-density lipoproteins and triglycerides, and raises high-density lipoprotein levels. Simvastatin is a lactone that is readily hydrolyzed in vivo to the corresponding β-hydroxyacid, and can be activated prior to use with NaOH in EtOH treatment. It is a synthetic analog of lovastatin (Cat. No. M2147).

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joost Besseling et al.
JAMA, 313(10), 1029-1036 (2015-03-11)
Familial hypercholesterolemia is characterized by impaired uptake of cholesterol in peripheral tissues, including the liver and the pancreas. In contrast, statins increase the cellular cholesterol uptake and are associated with increased risk for type 2 diabetes mellitus. We hypothesize that
Peter J Kirkpatrick et al.
The Lancet. Neurology, 13(7), 666-675 (2014-05-20)
The benefit of statins in patients with acute aneurysmal subarachnoid haemorrhage is unclear. We aimed to determine whether simvastatin 40 mg could improve the long-term outcome in patients with this disorder. In this international, multicentre, randomised, double-blind trial, we enrolled
Anders M Greve et al.
Stroke, 45(7), 1939-1946 (2014-06-07)
There are limited data on risk stratification of stroke in aortic stenosis. This study examined predictors of stroke in aortic stenosis, the prognostic implications of stroke, and how aortic valve replacement (AVR) with or without concomitant coronary artery bypass grafting
Jennifer G Robinson et al.
JAMA, 311(18), 1870-1882 (2014-05-16)
In phase 2 studies, evolocumab, a fully human monoclonal antibody to PCSK9, reduced LDL-C levels in patients receiving statin therapy. To evaluate the efficacy and tolerability of evolocumab when used in combination with a moderate- vs high-intensity statin. Phase 3
Laura Marin et al.
Expert opinion on drug delivery, 8(9), 1205-1220 (2011-05-28)
Chronic Obstructive Pulmonary Disease (COPD) is a severe disease that leads to a non-reversible obstruction of the small airways. The prevalence of this disease is rapidly increasing in developed countries, and in 2020 it has been predicted that this disease

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service