Skip to Content
Merck
All Photos(1)

Documents

T7080

Sigma-Aldrich

Tazarotene

≥98% (HPLC)

Synonym(s):

6-[(3,4-Dihydro-4,4-dimethyl-2H-1-benzothiopyran-6-yl)ethynyl]-3-pyridinecarboxylic acid ethyl ester, AGN-190168, ethyl 6-[2-(4,4-dimethylthiochroman-6-yl)ethynyl] nicotinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21NO2S
CAS Number:
Molecular Weight:
351.46
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to very faintly yellow

solubility

DMSO: >15 mg/mL

originator

Allergan

storage temp.

2-8°C

InChI

1S/C21H21NO2S/c1-4-24-20(23)16-7-9-17(22-14-16)8-5-15-6-10-19-18(13-15)21(2,3)11-12-25-19/h6-7,9-10,13-14H,4,11-12H2,1-3H3

InChI key

OGQICQVSFDPSEI-UHFFFAOYSA-N

Biochem/physiol Actions

Tazarotene induces the expression of tazarotene-induced gene 3 (TIG3), a tumor suppressor gene. It is a prodrug of tazarotenic acid, which specifically activates RARb and RARg, only weakly activates RARa, and is inactive at retinoid X receptors (RXRs). In psoriasis, tazarotene normalizes abnormal keratinocyte differentiation and reduces their hyperproliferation.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Allergan. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Guenther
Journal of the American Academy of Dermatology, 43(2 Pt 3), S36-S42 (2000-07-18)
In the hope of increasing efficacy and improving safety, several combination regimens involving tazarotene gel have been explored for the treatment of plaque psoriasis. A number of large-scale studies have shown that the adjunctive use of a mid-potency or high-potency
Yu-Chen Huang et al.
Lasers in surgery and medicine, 45(2), 102-107 (2013-02-21)
Nail psoriasis is difficult to treat. The efficacy of pulsed dye laser (PDL) therapy for nail psoriasis has been reported in non-placebo controlled studies. To evaluate the efficacy and safety of PDL with topical retinoid in the treatment of nail
Jorge Viera-Vera et al.
Biomolecules, 9(12) (2019-12-19)
Almost every organism has the ability of repairing damaged tissues or replacing lost and worn out body parts, nevertheless the degree of the response substantially differs between each species. Adult sea cucumbers from the Holothuria glaberrima species can eviscerate various
Mark A Hyde et al.
Archives of dermatology, 148(5), 592-596 (2012-03-21)
To determine if the complete response rates of lentigo maligna (LM) to imiquimod, 5%, cream can be improved by the addition of a topical retinoid. Prospective randomized study of patients treated with imiquimod alone vs imiquimod plus a topical retinoid
D D Tang-Liu et al.
Clinical pharmacokinetics, 37(4), 273-287 (1999-12-20)
Tazarotene (AGN 190168) is a new acetylenic retinoid which is effective for the topical treatment of patients with stable plaque psoriasis and mild to moderate acne vulgaris. Topical gel application provides direct delivery of tazarotene into the skin. At 10

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service