Skip to Content
Merck
All Photos(1)

Key Documents

1500218

USP

Paroxetine hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Paroxetine hydrochloride hemihydrate, (3S-trans)-3-[(1,3-Benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)piperidine hydrochloride hemihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20FNO3 · HCl · .5 H2O
CAS Number:
Molecular Weight:
374.83
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

paroxetine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O.Cl[H].Fc1ccc(cc1)[C@@H]2CCNC[C@H]2COc3ccc4OCOc4c3

InChI

1S/C19H20FNO3.ClH.H2O/c20-15-3-1-13(2-4-15)17-7-8-21-10-14(17)11-22-16-5-6-18-19(9-16)24-12-23-18;;/h1-6,9,14,17,21H,7-8,10-12H2;1H;1H2/t14-,17-;;/m0../s1

InChI key

QRQSGFFISBKLMZ-YHOFXEKLSA-N

Gene Information

human ... SLC6A4(6532)

Looking for similar products? Visit Product Comparison Guide

General description

Paroxetine hydrochloride is a phenylpiperidine derivative which is selective serotonin reuptake inhibitor. The protein binding ability is high.[1] It has 2 forms, non-hygroscopic hemihydrate and hygroscopic anhydrate.[2] This drug is seen useful in treating premature ejaculation in men.[3]
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Paroxetine hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Biochem/physiol Actions

Paroxetine hydrochloride hemihydrate is one of the most potent and selective of the selective serotonin reuptake inhibitors (SSRI); antidepressant
Selective serotonin reuptake inhibitor; antidepressant

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C G McMahon et al.
International journal of impotence research, 11(5), 241-245 (1999-12-20)
To evaluate the efficacy of chronic and 'on demand' administration of paroxetine hydrochloride in the drug treatment of premature ejaculation (PE). Ninety-four normally potent men, aged 18-61 y (mean 39 y) with premature ejaculation were treated between January 1996 and
Perinatal outcome following third trimester exposure to paroxetine.
Costei, Adriana Moldovan, et al.
Archives of Pediatrics & Adolescent Medicine, 156.11, 1129-1132 (2002)
Solid-state forms of paroxetine hydrochloride.
Buxton, P. Christopher, Ian R. Lynch, and John M. Roe.
International Journal of Pharmaceutics, 42.1, 135-143 (1988)
E Di Girolamo et al.
Journal of the American College of Cardiology, 33(5), 1227-1230 (1999-04-08)
The purpose of the study was to determine whether the well tolerated serotonin reuptake inhibitor paroxetine hydrochloride could prevent vasovagal syncope in patients resistant to or intolerant of previous traditional therapies. Serotonergic mechanisms play a major role in the processes
Puja Panwar Hazari et al.
ChemMedChem, 9(2), 337-349 (2014-01-01)
A novel SiX-dipropargyl glycerol scaffold (X: H, F, or (18) F) was developed as a versatile prosthetic group that provides technical advantages for the preparation of dimeric radioligands based on silicon fluoride acceptor pre- or post-labeling with fluorine-18. Rapid conjugation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service