Skip to Content
Merck
All Photos(1)

Key Documents

1279804

USP

Fluoxetine hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(±)-N-Methyl-γ-[4-(trifluoromethyl)phenoxy]benzenepropanamine hydrochloride, LY-110,140 hydrochloride, Prozac®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18F3NO · HCl
CAS Number:
Molecular Weight:
345.79
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

fluoxetine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CNCCC(C1=CC=CC=C1)OC2=CC=C(C(F)(F)F)C=C2.[H]Cl

InChI

1S/C17H18F3NO.ClH/c1-21-12-11-16(13-5-3-2-4-6-13)22-15-9-7-14(8-10-15)17(18,19)20;/h2-10,16,21H,11-12H2,1H3;1H

InChI key

GIYXAJPCNFJEHY-UHFFFAOYSA-N

Gene Information

human ... SLC6A4(6532)

Looking for similar products? Visit Product Comparison Guide

Application

Fluoxetine hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Fluoxetine Tablets
  • Fluoxetine Delayed-Release Capsules
  • Olanzapine and Fluoxetine Capsules
  • Fluoxetine Oral Solution
  • Fluoxetine Capsules

Biochem/physiol Actions

Selective serotonin reuptake inhibitor; antidepressant.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Legal Information

Sold under license from Eli Lilly and Company.
Prozac is a registered trademark of Eli Lilly and Co.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R J Gonzalez-Rothi et al.
Chest, 107(6), 1763-1765 (1995-06-01)
We describe a patient who developed progressive dyspnea, lung infiltrates, and restrictive lung disease in association with the antidepressant fluoxetine hydrochloride (Prozac). The pathologic findings were consistent with hypersensitivity pneumonitis. An associated pulmonary phospholipidosis was also noted.
Paulina Jedynak et al.
Journal of psychiatric research, 56, 106-111 (2014-06-17)
The neurogenesis hypothesis of major depression has two main facets. One states that the illness results from decreased neurogenesis while the other claims that the very functioning of antidepressants depends on increased neurogenesis. In order to verify the latter, we
Sean C Godar et al.
Journal of psychiatric research, 56, 1-9 (2014-06-03)
The termination of serotonin (5-hydroxytryptamine, 5-HT) neurotransmission is regulated by its uptake by the 5-HT transporter (5-HTT), as well as its degradation by monoamine oxidase (MAO)-A. MAO-A deficiency results in a wide set of behavioral alterations, including perseverative behaviors and
Kristie T Ota et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 40(3), 711-718 (2014-09-03)
Major depressive disorder (MDD) is a debilitating and widespread illness that exerts significant personal and socioeconomic consequences. Recent genetic and brain-imaging studies suggest that bicaudal C homolog 1 gene (BICC1), which codes for an RNA-binding protein, may be associated with
Melody V Wu et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 39(8), 1861-1871 (2014-02-13)
Adult hippocampal neurogenesis is critically implicated in rodent models of stress and anxiety as well as behavioral effects of antidepressants. Whereas similar factors such as psychiatric disorder and antidepressant administration are correlated with hippocampal volume in humans, the relationship between

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service