Skip to Content
Merck
All Photos(1)

Key Documents

1440808

USP

Metoclopramide hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-Amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide monohydrochloride monohydrate, Metoclopramide monohydrochloride monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22ClN3O2 · HCl · H2O
CAS Number:
Molecular Weight:
354.27
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

metoclopramide

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Cl-].Clc1c(cc(c(c1)C(=O)NCCN(CC)CC)OC)N.O.[H+]

InChI

1S/C14H22ClN3O2.ClH.H2O/c1-4-18(5-2)7-6-17-14(19)10-8-11(15)12(16)9-13(10)20-3;;/h8-9H,4-7,16H2,1-3H3,(H,17,19);1H;1H2

InChI key

KJBLQGHJOCAOJP-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Metoclopramide hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monographs such as:
  • Metoclopramide Tablets
  • Metoclopramide Oral Solution
  • Metoclopramide Injection
  • Prazosin Hydrochloride

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Radwa A Mohamed et al.
The Journal of pharmacy and pharmacology, 65(11), 1607-1621 (2013-10-10)
The purpose of this study was to formulate and characterize metoclopramide solid lipid nanoparticles (MCP-SLNs) and incorporating it into suppository bases for treatment of nausea and vomiting, produced with chemotherapeutic agents, using one dose per day. MCP-SLNs was prepared using
Ray M Merrill et al.
BMC psychiatry, 13, 152-152 (2013-05-30)
To identify the incidence rate of spontaneous dyskinesia (SD) and tardive dyskinesia (TD) in a general population and to examine the association between dykinesia and potential risk factors (exposure to metoclopramide [MCP], antipsychotic drugs, and history of diabetes and psychoses).
Anastasios Koulaouzidis et al.
Current medical research and opinion, 29(9), 1171-1185 (2013-06-25)
The use of purging for bowel cleansing prior to small-bowel capsule endoscopy (SBCE) has now been established in clinical practice. Despite that, the number of incomplete SBCEs is still around 15-20%. To date, the use of prokinetics in SBCE -
Diana Egerton-Warburton et al.
Emergency medicine Australasia : EMA, 25(3), 207-212 (2013-06-14)
The study aims to determine if slow intravenous infusion of metoclopramide reduces the incidence of acute drug-induced akathisia (DIA) compared with intravenous bolus. A prospective, double-blind, double dummy trial of adult patients requiring intravenous metoclopramide in the ED. Participants were
Allen Lee
Journal of gastrointestinal surgery : official journal of the Society for Surgery of the Alimentary Tract, 17(9), 1553-1556 (2013-06-27)
Gastroparesis is a chronic motility disorder that leads to delayed gastric emptying and negatively impacts morbidity, mortality, and quality of life. This paper provides an overview of the pathophysiology leading to symptoms in gastroparesis, discusses tests for diagnosis, and examines

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service