Skip to Content
Merck
All Photos(1)

Key Documents

32473

Supelco

Metoclopramide

VETRANAL®, analytical standard

Synonym(s):

4-Amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide, Methoxychloroprocainamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22ClN3O2
CAS Number:
Molecular Weight:
299.80
Beilstein:
1884366
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCN(CC)CCNC(=O)c1cc(Cl)c(N)cc1OC

InChI

1S/C14H22ClN3O2/c1-4-18(5-2)7-6-17-14(19)10-8-11(15)12(16)9-13(10)20-3/h8-9H,4-7,16H2,1-3H3,(H,17,19)

InChI key

TTWJBBZEZQICBI-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Metoclopramide is a drug, which can be effective in the treatment of severe nausea and vomiting caused by the anticancer drug cis-platinum.
Metoclopramide is typically employed as an antiemetic drug or a gastrointestinal prokinetic drug in adults and children.

Application

Metoclopramide may be used as a reference standard for the determination of the analyte in serum samples using reversed-phase high-performance liquid chromatography (RP-HPLC) with electrochemical detection.
Metoclopramide may be used as an analytical reference standard for the determination of the analyte in:
  • Human plasma by liquid–liquid extraction (LLE) with dichloromethane followed by hydrophilic interaction chromatography-electrospray iionization-tandem mass spectrometry (HILIC-ESI-MS/MS) operating in selected reaction monitoring (SRM) mode of detection.

It may also be used as an internal standard for the determination of loratadine and desloratadine in:
  • Human plasma by high performance liquid chromatography (HPLC) coupled to MS/MS with multiple reaction monitoring (MRM) detection.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RP?HPLC method with electrochemical detection for the determination of metoclopramide in serum and its use in pharmacokinetic studies.
Lamparczyk H, et al.
Biomedical Chromatography, 15(8), 513-517 (2001)
David C Currow et al.
Journal of palliative medicine, 15(10), 1071-1075 (2012-08-01)
Understanding the performance of prescribed medications in day-to-day practice is important to minimize harm, maximize clinical benefits, and, eventually, better target the people who are most likely to benefit, especially in hospice/palliative care where there may be limited time to
Double-blind crossover study of the antiemetic efficacy of high-dose dexamethasone versus high-dose metoclopramide.
Aapro SM, et al.
Journal of Clinical Oncology, 2(5), 466-471 (1984)
Determination of metoclopramide in human plasma using hydrophilic interaction chromatography with tandem mass spectrometry.
Lee HW, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 877(18-19), 1716-1720 (2009)
Metoclopramide: pharmacology and clinical application.
Albibi R and McCALLUM RW
Annals of Internal Medicine, 98(1), 86-95 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service