Skip to Content
Merck
All Photos(1)

Key Documents

1356803

USP

Lactulose

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-O-β-D-Galactopyranosyl-D-fructofuranose, 4-O-β-D-Galactopyranosyl-D-fructose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Beilstein:
93773
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

lactulose

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@@H](O[C@@H]2[C@@H](CO)O[C@@](O)(CO)[C@H]2O)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C12H22O11/c13-1-4-6(16)7(17)8(18)11(21-4)22-9-5(2-14)23-12(20,3-15)10(9)19/h4-11,13-20H,1-3H2/t4-,5-,6+,7+,8-,9-,10+,11+,12+/m1/s1

InChI key

JCQLYHFGKNRPGE-WJONTELPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lactulose USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Lactulose Concentrate
  • Lactulose Solution

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J Zhao et al.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society, 26(6), 794-802 (2014-03-20)
Small intestinal bacterial overgrowth (SIBO) may be a cause of irritable bowel syndrome (IBS); however, current investigations have important limitations. We aimed to identify clinically relevant diagnostic criteria for SIBO based on lactulose hydrogen breath test (LHBT) alone and combined
A A Guerra-Ordaz et al.
Applied and environmental microbiology, 80(16), 4879-4886 (2014-06-08)
The potential of a prebiotic oligosaccharide lactulose, a probiotic strain of Lactobacillus plantarum, or their synbiotic combination to control postweaning colibacillosis in pigs was evaluated using an enterotoxigenic Escherichia coli (ETEC) K88 oral challenge. Seventy-two weanlings were fed four diets:
M R Clausen et al.
Drugs, 53(6), 930-942 (1997-06-01)
Lactulose is one of the most frequently utilised agents in the treatment of constipation and hepatic encephalopathy because of its efficacy and good safety profile. The key to understanding the possible modes of action by which lactulose achieves its therapeutic
Jun Fang et al.
Journal of pharmaceutical sciences, 103(10), 3235-3243 (2014-07-22)
Bacteria of micrometer size could accumulate in tumor based on enhanced permeability and retention (EPR) effect. We report here Lactobacillus casei (L. casei), a nonpathogenic facultatively anaerobic bacterium, preferentially accumulated in tumor tissues after intravenously (i.v.) injection; at 24 h
Parmjit S Panesar et al.
Biotechnology advances, 29(6), 940-948 (2011-08-23)
Lactulose a "bifidus factor" is composed of galactose and fructose, which can be produced by the isomerization of lactose. It is a prebiotic carbohydrate which stimulates the growth of health-promoting bacteria in the gastrointestinal tract, such as bifidobacteria and lactobacilli

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service