Skip to Content
Merck
All Photos(1)

Documents

A1206000

Lactose (anhydrous)

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Lactose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Beilstein:
93796
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

Agency

EP Reference Standard

API family

lactose

manufacturer/tradename

EDQM

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C=O

InChI

1S/C12H22O11/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12/h1,4-12,14-21H,2-3H2/t4-,5+,6+,7+,8-,9-,10+,11+,12-/m0/s1

InChI key

DKXNBNKWCZZMJT-JVCRWLNRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lactose (anhydrous) EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yujin Zhu et al.
The American journal of gastroenterology, 108(9), 1516-1525 (2013-08-07)
Bloating and distention are often attributed to dietary factors by patients with irritable bowel syndrome (IBS). This study examined the effects of gas production and visceral hypersensitivity on digestive symptoms after lactose ingestion in a population with lactase deficiency. IBS
Daniela Barile et al.
Current opinion in biotechnology, 24(2), 214-219 (2013-02-26)
Human milk oligosaccharides (HMO) are believed to have a range of biological activities beyond providing nutrition to the infant. Principal among these is that they may act as prebiotics. Prebiotics are dietary ingredients, usually oligosaccharides that provide a health benefit
Xiang Kou et al.
Advanced drug delivery reviews, 64(3), 220-232 (2011-11-30)
A dry powder inhaler (DPI) is a dosage form that consists of a powder formulation in a device which is designed to deliver an active ingredient to the respiratory tract. It has been extensively investigated over the past years and
Aasma Shaukat et al.
Annals of internal medicine, 152(12), 797-803 (2010-04-21)
Lactose intolerance resulting in gastrointestinal symptoms is a common health concern. Diagnosis and management of this condition remain unclear. To assess the maximum tolerable dose of lactose and interventions for reducing symptoms of lactose intolerance among persons with lactose intolerance
T He et al.
European journal of clinical investigation, 38(8), 541-547 (2008-06-25)
Lactose maldigestion and intolerance affect a large part of the world population. The underlying factors of lactose intolerance are not fully understood. In this review, the role of colonic metabolism is discussed, i.e. fermentation of lactose by the colonic microbiota

Articles

A new HPLC-RI method is proposed for the analysis of Lactose in milk using a Chromolith® NH2 column.

A new HPLC-RI method is proposed for the analysis of Lactose in milk using a Chromolith® NH2 column.

A new HPLC-RI method is proposed for the analysis of Lactose in milk using a Chromolith® NH2 column.

A new HPLC-RI method is proposed for the analysis of Lactose in milk using a Chromolith® NH2 column.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service