Skip to Content
Merck
All Photos(1)

Key Documents

1205003

USP

Diltiazem hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(+)-cis-Diltiazem hydrochloride, (2S,3S)-(+)-cis-3-Acetoxy-5-(2-dimethylaminoethyl)-2,3-dihydro-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one hydrochloride, CRD-401

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26N2O4S · HCl
CAS Number:
Molecular Weight:
450.98
Beilstein:
4228706
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

diltiazem

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.COc1ccc(cc1)[C@@H]2Sc3ccccc3N(CCN(C)C)C(=O)[C@@H]2OC(C)=O

InChI

1S/C22H26N2O4S.ClH/c1-15(25)28-20-21(16-9-11-17(27-4)12-10-16)29-19-8-6-5-7-18(19)24(22(20)26)14-13-23(2)3;/h5-12,20-21H,13-14H2,1-4H3;1H/t20-,21+;/m1./s1

InChI key

HDRXZJPWHTXQRI-BHDTVMLSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application


  • Exploring the Effectiveness of Carboxymethylated and Crosslinked Albizia procera Gum in Diltiazem Hydrochloride Matrix Tablets: A Comparative Analysis. This study investigates the potential of carboxymethylated and crosslinked Albizia procera gum for use in sustained-release matrix tablets containing Diltiazem hydrochloride, highlighting its viability in pharmaceutical applications (Mukherjee S, Khanam J, 2024).

  • Dietary gallic acid as an antioxidant: A review of its food industry applications, health benefits, bioavailability, nano-delivery systems, and drug interactions. While primarily focused on gallic acid, this review mentions Diltiazem hydrochloride in the context of drug interactions and its implications in food safety and health (Xiang Z et al., 2024).

  • Dual stimuli-responsive and sustained drug delivery NanoSensoGel formulation for prevention of cisplatin-induced ototoxicity. This research presents a novel NanoSensoGel that could be adapted for Diltiazem hydrochloride, enhancing drug delivery efficiency in clinical settings (Thakur NS et al., 2024).

  • A novel potentiometric sensor based on ZnO decorated polyaniline/coal nanocomposite for diltiazem determination. This study develops a new sensor for the precise measurement of Diltiazem levels in pharmaceutical formulations, which is crucial for quality control and regulatory compliance (El Sayed GA et al., 2023).

  • The Effect of Topical Nifedipine versus Diltiazem on the Acute Anal Fissure: A Randomized Clinical Trial. This clinical trial evaluates the effectiveness of Diltiazem hydrochloride in treating acute anal fissures, providing evidence of its beneficial applications in proctological disorders (Momayez Sanat Z et al., 2023).

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diltiazem hydrochloride.
B F McGraw
Drug intelligence & clinical pharmacy, 16(5), 366-370 (1982-05-01)
[Diltiazem hydrochloride: synthetic and pharmacological studies and development].
K Abe et al.
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 108(8), 716-732 (1988-08-01)
K Adibkia et al.
Drug research, 64(4), 203-207 (2013-09-13)
The aim of this study was to design and evaluate diltiazem hydrochloride controlled porosity osmotic pump (CPOP) system employing a new coating technique named the micro suspension coating method. In this method the fast dissolving pore formers were suspended in
Takeshi Ogura et al.
Acta neurochirurgica. Supplement, 120, 47-50 (2014-11-05)
This study evaluated the effect of diltiazem, a calcium antagonist, on sympathetic activity in patients with aneurysmal subarachnoid hemorrhage (SAH) during the hyperacute stage. Of patients with aneurysmal SAH who underwent aneurysm repair between August 2008 and June 2011, 119
B J McAuley et al.
Pharmacotherapy, 2(3), 121-133 (1982-05-01)
Diltiazem, a calcium channel blocking agent, has potent cardiovascular effects that are directly related to its influence on vascular smooth muscle, ventricular myocardium, and specialized conducting tissue. It causes coronary and peripheral vasodilation, has a negative chronotropic and dromotropic effect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service