Skip to Content
Merck
All Photos(1)

Key Documents

C2615

Sigma-Aldrich

Chromanol 293B

≥98% (HPLC), powder

Synonym(s):

trans-N-[6-Cyano-3,4-dihydro-3-hydroxy-2,2-dimethyl-2H-1-benzopyran-4-yl]-N-methyl-ethanesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20N2O4S
CAS Number:
Molecular Weight:
324.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white

solubility

DMSO: 18 mg/mL

storage temp.

−20°C

SMILES string

CCS(=O)(=O)N(C)[C@@H]1[C@@H](O)C(C)(C)Oc2ccc(cc12)C#N

InChI

1S/C15H20N2O4S/c1-5-22(19,20)17(4)13-11-8-10(9-16)6-7-12(11)21-15(2,3)14(13)18/h6-8,13-14,18H,5H2,1-4H3/t13-,14+/m0/s1

InChI key

HVSJHHXUORMCGK-UONOGXRCSA-N

General description

Chromanol 293B enantiomer is a potent inhibitor of potassium channel protein (KvLQT1).[1] In human atrial myocytes, chromanol 293B inhibits repolarization potassium currents.[2] Chromanol 293B improves glucose-stimulated insulin secretion (GSIS) in pancreas by modulating potassium voltage-gated channel (KCNQ1).[3]

Application

Chromanol 293B has been used in the inhibition of calcium and cyclic adenosine monophosphate (cAMP)-activated potassium channels in human epithelial cell lines.[4][5] Chromanol 293B has been used in patch-clamp electrophysiology studies in cardiomyocytes.[6]

Biochem/physiol Actions

Blocker of the slow delayed rectifier K+ current via KCNQ1 channels

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereoselective interactions of the enantiomers of chromanol 293B with human voltage-gated potassium channels
Yang IC, et al.
Journal of Pharmacology and Experimental Therapeutics, 294(3), 955-962 (2000)
CFTR-mediated anion secretion across intestinal epithelium-like Caco-2 monolayer under PTH stimulation is dependent on intermediate conductance K+ channels
Jantarajit W, et al.
American Journal of Physiology. Cell Physiology, 313(1), C118-C129 (2017)
J E Matos et al.
Acta physiologica (Oxford, England), 189(3), 251-258 (2007-02-20)
Colonic crypts are the site of Cl- secretion. Basolateral K+ channels provide the driving force for luminal cystic fibrosis transmembrane regulator-mediated Cl- exit. Relevant colonic epithelial K+ channels are the intermediate conductance Ca2+-activated K(Ca)3.1 (SK4) channel and the cAMP-activated K(V)7.1
A Varro et al.
The Journal of physiology, 523 Pt 1, 67-81 (2000-02-16)
1. The relative contributions of the rapid and slow components of the delayed rectifier potassium current (IKr and IKs, respectively) to dog cardiac action potential configuration were compared in ventricular myocytes and in multicellular right ventricular papillary muscle and Purkinje
Effects of chromanol 293B on transient outward and ultra-rapid delayed rectifier potassium currents in human atrial myocytes
Du XL, et al.
Journal of Molecular and Cellular Cardiology, 35(3), 293-300 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service