Skip to Content
Merck
All Photos(1)

Key Documents

1131009

USP

Cholecalciferol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(+)-Vitamin D3, 7-Dehydrocholesterol activated, Activated 7-dehydrocholesterol, Calciol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O
CAS Number:
Molecular Weight:
384.64
Beilstein:
2339331
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

cholecalciferol

manufacturer/tradename

USP

mp

83-86 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CC(C)CCC[C@@H](C)[C@@]1([H])CC[C@@]([C@]1(C)CCC/2)([H])C2=C\C=C(C[C@@H](O)CC3)/C3=C

InChI

1S/C27H44O/c1-19(2)8-6-9-21(4)25-15-16-26-22(10-7-17-27(25,26)5)12-13-23-18-24(28)14-11-20(23)3/h12-13,19,21,24-26,28H,3,6-11,14-18H2,1-2,4-5H3/b22-12+,23-13-/t21-,24+,25-,26+,27-/m1/s1

InChI key

QYSXJUFSXHHAJI-YRZJJWOYSA-N

Gene Information

human ... VDR(7421)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cholecalciferol USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Cholecalciferol Solution
  • Cholecalciferol Capsules
  • Cholecalciferol Tablets
  • Cholecalciferol Chewable Gels
  • Calcium with Vitamin D Tablets
  • Oil-Soluble Vitamins Preparation

Biochem/physiol Actions

Vitamin D acts through a receptor that is a member of the ligand-dependent transcription factor superfamily. Modulates the proliferation and differentiation of both normal and cancer cells. Has antiproliferative and antimetastatic effects on breast, colon, and prostate cancer cells. Activated vitamin D receptors in intestine and bone maintain calcium absorbance and homeostasis.
Vitamin D acts through a receptor that modulates the proliferation and differentiation of both normal and cancer cells.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - STOT RE 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cholecalciferol Solution
United States Pharmacopeia, 987-987 (2013)
Cholecalciferol
United States Pharmacopeia, 44(6), 985-985 (2020)
Cholecalciferol Tablets
United States Pharmacopeia, 43(2), 988-988 (2018)
Robert Scragg et al.
Hypertension (Dallas, Tex. : 1979), 64(4), 725-730 (2014-07-02)
Previous randomized controlled trials of vitamin D supplementation and blood pressure (BP) mainly have given vitamin D for short periods (<6 months) or at low doses (400 IU per day). This study aims to determine whether long-term high-dose vitamin D
Sagar U Nigwekar et al.
American journal of kidney diseases : the official journal of the National Kidney Foundation, 60(1), 139-156 (2012-05-09)
The development of chronic kidney disease (CKD) is accompanied by a progressive decrease in the ability to produce 1,25-dihydroxyvitamin D. Pharmacological replacement with active vitamin D therefore has been a cornerstone of secondary hyperparathyroidism therapy in the end-stage renal disease

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service