Skip to Content
Merck
All Photos(1)

Key Documents

PHR1238

Supelco

Ergocalciferol (Vitamin D2)

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Ergocalciferol, Calciferol, Ercalciol, Ergosterol irradiated, Irradiated ergosterol, Vitamin D2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H44O
CAS Number:
Molecular Weight:
396.65
Beilstein:
1916682
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 788
traceable to Ph. Eur. E0900000
traceable to USP 1239005

API family

ergocalciferol

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

114-118 °C (lit.)

application(s)

clinical testing

format

neat

storage temp.

-10 to -25°C

SMILES string

CC(C)[C@@H](C)/C=C/[C@@H](C)[C@@]1([H])CC[C@@]([C@]1(C)CCC/2)([H])C2=C\C=C(C[C@@H](O)CC3)/C3=C

InChI

1S/C28H44O/c1-19(2)20(3)9-10-22(5)26-15-16-27-23(8-7-17-28(26,27)6)12-13-24-18-25(29)14-11-21(24)4/h9-10,12-13,19-20,22,25-27,29H,4,7-8,11,14-18H2,1-3,5-6H3/b10-9+,23-12+,24-13-/t20-,22+,25-,26+,27-,28+/m0/s1

InChI key

MECHNRXZTMCUDQ-RKHKHRCZSA-N

Gene Information

human ... VDR(7421)

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Ergocalciferol (Vitamin D2) may be used as a pharmaceutical reference standard for the determination of the analyte in serum samples, cosmetics and pharmaceutical formulations by various chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB7854 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - STOT RE 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A RAPID METHOD FOR DETERMINATION OF VITAMINS D2 AND D3 IN PHARMACEUTICAL PREPARATIONS BY HPLC
Sarioglu K, et al.
Journal of Liquid Chromatography and Related Technologies, 24(7), 973-982 (2001)
Is supercritical fluid chromatography hyphenated to mass spectrometry suitable for the quality control of vitamin D3 oily formulations?
Andri B. et al.
Journal of Chromatography A, 1515(7), 209-217 (2017)
Determination of vitamins A, D, E, and K in human and bovine serum, and ?-carotene and vitamin A palmitate in cosmetic and pharmaceutical products, by isocratic HPLC
Wang LH and Huang SH
Chromatographia, 55(5-6), 289-296 (2002)
H A Bischoff-Ferrari et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 19(2), 265-269 (2004-02-19)
Intracellular 1,25-dihydroxyvitamin D receptor (VDR) is expressed in human skeletal muscle tissue. However, it is unknown whether VDR expression in vivo is related to age or vitamin D status, or whether VDR expression differs between skeletal muscle groups. We investigated
Amanda J Samanek et al.
The Medical journal of Australia, 184(7), 338-341 (2006-04-06)
To examine the influence of geographical and seasonal factors on duration of solar ultraviolet (UV) radiation exposure of skin to produce recommended vitamin D levels without producing erythema. An ecological study using daily Ultraviolet Index (UVI) data collected in major

Protocols

Ascentis Express C18 is suitable for routine analysis of fat-soluble vitamins D2 and D3, providing optimized chromatograms.

Ascentis Express C18 is suitable for routine analysis of fat-soluble vitamins D2 and D3, providing optimized chromatograms.

Ascentis Express C18 is suitable for routine analysis of fat-soluble vitamins D2 and D3, providing optimized chromatograms.

Separation of Ergocalciferol (D2), analytical standard; Cholecalciferol (D3), analytical standard

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service