Skip to Content
Merck
All Photos(1)

Key Documents

RAD130S

radiello Passive Sampler Starter Kit

BTEX/VOC, CS2 Desorption

Synonym(s):

radiello Starter Kits

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

Product Name

radiello BTEX/VOC Starter Kit, CS2 Desorption,

Agency

EPA TO-14A, TO-15, TO-17
NIOSH 1500, 1501, 2549
OSHA 111, 1002, 1004

packaging

pkg of 1 kit

manufacturer/tradename

Radiello

technique(s)

passive air sampling: suitable

matrix

SS net (100 mesh, 5.8 mm diam.), activated with activated charcoal (30-50 mesh)

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for sampling BTEX / VOCs (CS <SUB>2</SUB> Desorption)

Looking for similar products? Visit Product Comparison Guide

General description


Each RAD130S Starter Kit includes:
Two radiello cartridge adsorbents (RAD130) for VOCs - CS2 Desorption (Note: 1 x cartridge for sampling; and1 x for blank)
1 x Triangular support plate (RAD121)
1 x vertical adapter for personal sampling (RAD122)
1 x white diffusive body (RAD120)
Detailed sampling/analysis instructions
Radiello Starter Kits are ideal for introducing you to the product and include everything you need to take one complete sample. No additional parts are required. Each kit includes the following items:

Each RAD130S Starter Kit Includes:
Two radiello cartridge adsorbents - Solvent Desorption Note: 1 x cartridge for sampling; and 1 x for blank)
1 x Triangular support plate (RAD121)
1 x vertical adapter for personal sampling (RAD122)
1 x white diffusive body (RAD120)
1 x barcode label (RAD190)
Detailed sampling/analysis instructions


Each RAD141S and 145S Starter Kit Includes:
Two radiello cartridge adsorbents - Thermal Desorption (Note: 1 x cartridge for sampling; and 1 x for blank)
1 x Triangular support plate (RAD121)
1 x vertical adapter for personal sampling (RAD122)
1 x yellow diffusive body (RAD1202)
1 x barcode label (RAD190)
Detailed sampling/analysis instructions

Application

A radiello diffusive sampler may be used for thermal desorption during passive sampling, in an comparative study done to evaluate the relative performance of active and passive sampling methods in order to analyse the volatile organic compounds (VOCs) in ambient air. A radiello cartridge packed with Carbograph 4 may also be used in diffusive sampling to determine time-weighted average volatile organic compound (VOC) concentration in occupational hygiene and environmental air monitoring. A radiello diffusive sampler may also be used to monitor the cadaveric VOCs released by decomposing pig carcasses in three biotopes (crop field, forest, urban site).

Packaging

Kit includes: 2 RAD130 adsorbing cartridges and barcode labels, 1 RAD120 white diffusive body, 1 RAD121 triangular support plate, 1 instruction sheet

Legal Information

radiello is a trademark of Institi Clinici Scientifici Maugeri

Disclaimer

*Note: This product is not officially specified in the method, it may be suitable for use for sampling compound(s) listed in the method*

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Passive Sampling of Atmospheric Organic Contaminants
Esteve-Turrillas, F.A., et al.
Comprehensive Sampling and Sample Preperation, 1, 201-222 (2012)
Volatile organic compounds in air
Methods for the Determination of Hazardous Substances, 1-17 (1997)
Study of the dispersion of VOCs emitted by a municipal solid waste landfill
Chiriac, R., et al.
Atmospheric Environment, 43 (11), 1926-1931 (2009)
Isabel M Morales Terrés et al.
Journal of environmental management, 91(12), 2754-2762 (2010-09-03)
This paper describes a novel methodology for evaluating the extent to which petrol stations affect their surroundings. The method is based on the fact that the ratio of the concentrations of aliphatic and aromatic hydrocarbon pollutants in the air of
C Minoia et al.
The Science of the total environment, 192(2), 163-182 (1996-12-02)
A study using individual dosimetry to evaluate the daily inhaled dose of sixteen aromatic and aliphatic hydrocarbons in three groups of primary school children, living in three Italian towns with 50,000 inhabitants or less, (Treviglio-Lombardy; Poggibonsi-Tuscany; Valenza-Piedmont) is presented. The

Articles

Learn the how the diffusive air sampler works to see if it’s the right fit for you.

Learn the how the diffusive air sampler works to see if it’s the right fit for you.

Protocols

Learn how to assemble the supporting plate of your radiello air sampler in the lab before the sampling campaign to save time in the field.

Related Content

Complete offer for petrochemical testing including environmental and industrial hygiene monitoring solutions.

Complete offer for petrochemical testing including environmental and industrial hygiene monitoring solutions.

Complete offer for petrochemical testing including environmental and industrial hygiene monitoring solutions.

Complete offer for petrochemical testing including environmental and industrial hygiene monitoring solutions.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service