Skip to Content
Merck
All Photos(1)

Key Documents

PHL89514

Campesterol

phyproof® Reference Substance

Synonym(s):

24α-Methyl-5-cholesten-3β-ol, 24(R)-Ergost-5-en-3β-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H48O
CAS Number:
Molecular Weight:
400.68
Beilstein:
3216975
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

biological source

Glycine max (soybean)

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (GC)

form

crystals

manufacturer/tradename

PhytoLab

storage temp.

2-8°C

SMILES string

[H][C@@]12CC=C3C[C@@H](O)CC[C@]3(C)[C@@]1([H])CC[C@]4(C)[C@H](CC[C@@]24[H])[C@H](C)CC[C@H](C)C(C)C

InChI

1S/C28H48O/c1-18(2)19(3)7-8-20(4)24-11-12-25-23-10-9-21-17-22(29)13-15-27(21,5)26(23)14-16-28(24,25)6/h9,18-20,22-26,29H,7-8,10-17H2,1-6H3/t19-,20+,22-,23-,24+,25-,26-,27-,28+/m0/s1

InChI key

SGNBVLSWZMBQTH-ZRUUVFCLSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG.

Campesterol is a major phytosterol having a core skeleton similar to cholesterol but with a methyl side chain at C24. It is used in the management of hypercholesterolemia, besides having antioxidant, anti-inflammatory, and anti-cancer properties.

Application

This primary reference standard can also be used as follows:

  • Development of two high-performance liquid chromatography (HPLC)-diode array ultraviolet/visible detection (DAD-UV/VIS) and liquid chromatography-mass spectrometry (LC-MS) methods to determine campesterol, stigmasterol, and daucosterol from the herbal samples of Artemisia apiacea
  • Multi-residue determination of β-sitosterol, campesterol, and stigmasterol in rat plasma samples by liquid chromatography-atmospheric pressure chemical ionization-tandem mass spectrometry (LC-APCI-MS/MS) method
  • Ultrasound-assisted extraction (UAE) of campesterol, β-sitosterol, and stigmasterol from passion fruit seed oil samples for their determination by gas chromatography-flame ionization detection (GC-FID)
  • Analysis of 119 vegetable oil samples from 7 different varieties to detect and determine squalene and four sterols using a developed and validated non-destructive method based on proton nuclear magnetic resonance (1H NMR) spectroscopy combined with partial least squares (PLS) method
  • Multi-residue analysis of isoquercitrin, campesterol, emodin 8-O-β-D-glucopyranoside, and quercetin in dried stem and flower extracts of Reynoutria sachalinensis by high-performance liquid chromatography coupled with diode-array detection

Other Notes

This compound is commonly found in plants of the genus: ginkgo hedera lycium sambucus silybum triticum

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service