Skip to Content
Merck
All Photos(1)

Documents

85451

Sigma-Aldrich

β-Sitosterol

≥70%

Synonym(s):

α-Dihydrofucosterol, 22,23-Dihydrostigmasterol, 24α-Ethylcholesterol, 5-Stigmasten-3β-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H50O
CAS Number:
Molecular Weight:
414.71
Beilstein:
1916165
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

biological source

plant

Assay

≥70%

form

powder

impurities

campesterol and β-sitostanol, residual

mp

136-140 °C (lit.)

functional group

hydroxyl

SMILES string

O[C@H](C1)CC[C@@]2(C)C1=CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4([H])[C@H](C)CC[C@H](C(C)C)CC

InChI

1S/C29H50O/c1-7-21(19(2)3)9-8-20(4)25-12-13-26-24-11-10-22-18-23(30)14-16-28(22,5)27(24)15-17-29(25,26)6/h10,19-21,23-27,30H,7-9,11-18H2,1-6H3/t20-,21-,23+,24+,25-,26+,27+,28+,29-/m1/s1

InChI key

KZJWDPNRJALLNS-VJSFXXLFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

β-Sitosterol was used to supplement diet of mice to study its effect on body mass accumulation.

Biochem/physiol Actions

β-Sitosterol is a plant sterol that has hypocholesterolemic activity. Only 5% of β-Sitosterol is absorbed by humans. It is hypothesized that the cholesterol-lowering activity is by competing cholesterol for uptake by the cells or by interfering with the esterification of cholesterol. β-Sitosterol is reported to reduce the incidence of colon tumors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M Sugano et al.
The Journal of nutrition, 107(11), 2011-2019 (1977-11-01)
The hypocholesterolemic activity of beta-sitosterol and its hydrogenated product, beta-sitostanol (dihydrositosterol or stigmastanol) has been compared in young male rats. When cholesterol was included in the diet, sitostanol consistently exhibited significantly greater hypocholesterolemic activity than sitosterol. There were no apparent
Yoshihide Yamanashi et al.
The Journal of pharmacology and experimental therapeutics, 320(2), 559-564 (2006-12-01)
Previous in vivo studies including those with knockout mice suggested that Niemann-Pick C1-like 1 (NPC1L1) plays an essential role in the intestinal absorption of cholesterol. To characterize the mechanism of cholesterol uptake mediated by NPC1L1, an in vitro system reflecting
Pauline Seguinot et al.
Frontiers in microbiology, 11, 305-305 (2020-03-19)
Non-Saccharomyces yeasts are currently widely used in winemaking to enhance aroma profile diversity among wines. The use of Metschnikowia pulcherrima in sequential inoculation with S. cerevisiae was compared to the inoculation of a pure culture of S. cerevisiae. Moreover, various
P P Nair et al.
The American journal of clinical nutrition, 40(4 Suppl), 927-930 (1984-10-01)
Cholesterol and fat are implicated as dietary factors enhancing the risk for colon carcinogenesis. Plant sterols such as beta-sitosterol when added to diets of experimental animals treated with colon carcinogens reduce tumor yields and counteract the proliferative changes associated with
F J Field et al.
Journal of lipid research, 24(4), 409-417 (1983-04-01)
Rabbits were fed either 10% coconut oil, 10% coconut oil and 1% beta-sitosterol, 10% coconut oil and 1% cholesterol, or 10% coconut oil and 1% beta-sitosterol plus 1% cholesterol for 4 weeks. Microsomal membranes from intestines of animals fed the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service