Skip to Content
Merck
All Photos(1)

Documents

CRM43234

Supelco

Methiocarb

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Mercaptodimethur

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15NO2S
CAS Number:
Molecular Weight:
225.31
Beilstein:
1881431
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

2-30°C

SMILES string

CNC(=O)Oc1cc(C)c(SC)c(C)c1

InChI

1S/C11H15NO2S/c1-7-5-9(14-11(13)12-3)6-8(2)10(7)15-4/h5-6H,1-4H3,(H,12,13)

InChI key

YFBPRJGDJKVWAH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Methiocarb is a synthetic carbamate pesticide that shows broad biological activity. It is widely used for its insecticidal and molluscicidal activity against snails, rice insects, and also on fruit crops for its bird-repelling nature.
As per the commission implementing regulation (EU) 2019/1606 of 27th September 2019, methiocarb (expressed as a sum of methiocarb, methiocarb sulfone, and methiocarb sulfoxide) is not approved for use in the European Union. A maximum residue limit (MRLs) of 0.01 to 0.1 mg/kg has been set for the presence of methiocarb in various products of plant and animal origin as per Reg (EU) 2021/155. Methiocarb is to be monitored in the Multiannual Control Programme for Pesticides Residues (MACP), run within the EU and EFTA in/on products of plant origin.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
The methiocarb CRM may also find the following uses:

  • Determination of methiocarb in water sample using boron-doped diamond electrode (BDDE) and differential pulse voltammetry
  • Development of a method to simultaneously measure methiocarb and its metabolites in five food products of animal origin (milk, chicken, pork, beef, and eggs) by liquid chromatography-tandem mass spectrometry(LC-MS/MS)
  • Extraction and quantification of six carbamate pesticide residues in water, fruit, and vegetable samples using dispersive liquid-liquid microextraction and high-performance liquid chromatography
  • Simultaneous analysis of trace carbamate pesticide residues in tomato samples using graphene-based magnetic nanoparticles for their magnetic solid phase extraction and high-performance liquid chromatographic determination
  • Pre-concentration of 13 pesticide residues using a silica-based MSU-1 mesoporous material and their subsequent determination by UPLC-MS/MS in environmental water samples
  • Multi-residue determination of 200 pesticides and their metabolites in honeybee samples using liquid and gas chromatography coupled to tandem mass spectrometry

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Kharbouche et al.
Talanta, 199, 612-619 (2019-04-07)
This paper describes the synthesis of a silica based MSU-1 mesoporous solid and its application as sorbent in solid-phase extraction to pre-concentrate thirteen pesticides of low-high polarity (methomyl, cymoxanil, carbofuran, monolinuron, isoproturon, methidathion, methiocarb, malathion, phosalone, diazinon, penconazole, neburon and
Na Li et al.
Talanta, 141, 212-219 (2015-05-13)
Graphene-based magnetic nanoparticles, comprising zero-valent iron, iron oxide-oxyhydroxide and graphene, were prepared through a simple one-step synthesis method, and subsequently applied to magnetic solid-phase extraction for the determination of trace carbamate pesticides in tomatoes coupled with high performance liquid chromatography.
Determination of methiocarb pesticide using differential pulse voltammetry with a boron-doped diamond electrode
Chylkova J, et al.
Analytical Methods : Advancing Methods and Applications, 7(11), 4671-4677 (2015)
Determination of carbamate insecticides in water, fruit, and vegetables by ultrasound-assisted dispersive liquid-liquid microextraction and high-performance liquid chromatography
Kongphonprom K and Burakham R
Analytical Letters, 49(6), 753-767 (2016)
Md Musfiqur Rahman et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1060, 387-394 (2017-07-01)
A simultaneous analytical method was developed for the determination of methiocarb and its metabolites, methiocarb sulfoxide and methiocarb sulfone, in five livestock products (chicken, pork, beef, table egg, and milk) using liquid chromatography-tandem mass spectrometry. Due to the rapid degradation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service