Skip to Content
Merck
All Photos(1)

Key Documents

36184

Supelco

Oxamyl

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13N3O3S
CAS Number:
Molecular Weight:
219.26
Beilstein:
2212753
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water (Karl Fischer)

mp

95-101 °C

suitability

passes test for identity (NMR)

application(s)

agriculture
environmental

format

neat

SMILES string

CNC(=O)O\N=C(/SC)C(=O)N(C)C

InChI

1S/C7H13N3O3S/c1-8-7(12)13-9-5(14-4)6(11)10(2)3/h1-4H3,(H,8,12)/b9-5-

InChI key

KZAUOCCYDRDERY-UITAMQMPSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gerardo A Anguiano et al.
Environmental toxicology, 25(4), 327-332 (2009-05-19)
Acetylcholinesterase (AChE) activity has been used to test the exposure of mollusk bivalves to pesticides and other pollutants. The Pacific oyster Crassostrea gigas is a species with a worldwide distribution, and it has a high commercial value. The use of
Emmanuel A Tzortzakakis et al.
Pest management science, 59(12), 1311-1320 (2003-12-12)
The root galling index and the densities of eggs in roots and juveniles in soil of the root-knot nematode Meloidogyne javanica (Treub) Chitwood on tomato, and the effect of these on crop yield were assessed in greenhouse experiments applying various
F Lamberti et al.
Mededelingen (Rijksuniversiteit te Gent. Fakulteit van de Landbouwkundige en Toegepaste Biologische Wetenschappen), 67(3), 699-702 (2003-04-17)
Bunches of 25 g of tomato galled roots containing ca. 270 eggs and juveniles/g of Meloidogyne incognita were buried on 29 September 2001 in gauze bags at the depth of 15, 30 and 45 cm in a sandy loam. The
M L Parker et al.
Archives of environmental contamination and toxicology, 39(2), 233-242 (2000-06-28)
The concept of B-esterase buffering against anti-cholinesterase (ChE) insecticide toxicity has been extensively researched in mammalian species. Presumably due to relatively low levels of anti-ChE detoxifying enzyme activity in birds, however, avian species are often more susceptible to the toxic
E Grauer et al.
Toxicology, 242(1-3), 1-6 (2007-10-13)
Severe poisoning by inhibitors of cholinesterase (ChE) enzymes is often associated with prolonged central or peripheral neuronal damage. Oxotremorine is a cholinergic agonist known to induce acute hypothermia. Central and peripheral cholinergic signaling is involved in the induction of hypothermia

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service