Skip to Content
Merck
All Photos(1)

Key Documents

73003AST

Supelco

Astec® CHIRALDEX A-TA Capillary GC Column

L × I.D. 30 m × 0.25 mm, df 0.12 μm

Synonym(s):

GC column, chiral, alpha-dextran

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115710
eCl@ss:
32110502
NACRES:
SB.54

material

fused silica

description

GC capillary column

packaging

pkg of 1 ea

parameter

-10-180 °C temperature (isothermal or programmed)

Beta value

500

df

0.12 μm

technique(s)

gas chromatography (GC): suitable

L × I.D.

30 m × 0.25 mm

matrix active group

non-bonded; 2,6-di-O-pentyl-3-trifluoroacetyl derivative of α-cyclodextrin phase

application(s)

agriculture
chemicals and industrial polymers
cleaning products
clinical
cosmetics
environmental
flavors and fragrances
food and beverages
forensics and toxicology
life science and biopharma
personal care
pharmaceutical (small molecule)

column type

capillary chiral

separation technique

chiral

Looking for similar products? Visit Product Comparison Guide

General description

Incorporates a phase consisting of a 2,6-di-O-pentyl-3-trifluoroacetyl derivative of α-cyclodextrin. This phase exhibits high selectivity for oxygen-containing analytes in the form of alcohols, ketones, acids, aldehydes and lactones. It is also highly selective for halogenated compounds.

Chem/Phys Resistance

Temp. Limits:
  • -10 °C to 180 °C, isothermal and programmed

Other Notes

We offer a variety of chromatography accessories including analytical syringes

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly enantioselective 1,4-conjugate addition of dialkylzinc to a,?-unsaturated lactone catalyzed by diphosphite?copper complexes
Liang, Liang, et al.
Tetrahedron Letters, 44 (38), 7217-7220 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service