Skip to Content
Merck
All Photos(1)

Key Documents

11249738001

Roche

Phytohemagglutinin-L (PHA-L)

from Phaseolus vulgaris

Synonym(s):

PHA-L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
Pricing and availability is not currently available.

Quality Level

sterility

non-sterile; 0.2 μm filtered

Assay

>95% (SDS-PAGE)

form

lyophilized (clear, colorless solution after reconstitution)

packaging

pkg of 5 mg

manufacturer/tradename

Roche

application(s)

hematology

storage temp.

2-8°C

Related Categories

General description

PHA-L is purified from Phaseolus vulgaris (red kidney bean) by standard chromatographic techniques.
Phytohemagglutinin (PHA), the lectin extract from the red kidney bean, consists of only L-type subunits (isolectin L4, "leuko-agglutinin"). It is an N-acetylgalactosamine/galactose sugar-specific lectin. PHA consists of five isolectins (L4E0, L3E1, L2E2, L1E3, L0E4) each being a tetramer held together by noncovalent forces. The subunits L (leukocyte reactive) have a high affinity for lymphocyte surface receptors, but little for those of erythrocytes, and are responsible for the mitogenic properties of the isolectins. E (erythrocyte reactive) is responsible for the erythrocyte-agglutinating properties.[1]

Application

Phytohemagglutinin-L (PHA-L) consists of only L-type subunits (isolectin L4, "leuko-agglutinin"), and is well suited for high-efficiency induction and the functional analysis of T-lymphocytes.[2] It is highly purified and tested for high-efficiency stimulation of human lymphocytes. It is also used as positive controls for peripheral blood mononuclear cells (PBMC) stimulation.[3][4]

Biochem/physiol Actions

Phytohemagglutinin (PHA) has mitogenic and potent cell agglutinating activities. It binds to T-cell membranes and stimulates cell division and metabolic activity. PHA has the ability to stimulate close contacts between cell membranes.[1]

Specifications

Biological activity: <5μg/ml for maximal stimulation of DNA synthesis (BrdU incorporation) with human peripheral blood lymphocytes

Sequence

PHA-L is a tetramer of 4 L-type subunits held together by noncovalent forces.

Preparation Note

Working concentration: 1 to 5 μg/ml
Approximately 1 to 5 μg/ml for the stimulation of human peripheral blood lymphocytes
Storage conditions (working solution): -15 to -25 °C
The reconstituted solution is stable at -15 to -25 °C.
Note: Store the reconstituted solution in aliquots at -15 to -25 °C.
Avoid repeated freezing and thawing.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian-Fei Hu et al.
Frontiers in immunology, 13, 958960-958960 (2022-08-23)
Chimeric antigen receptor-engineered T cell (CAR-T) therapy has shown promising potential for anti-cancer treatment. However, for pancreatic ductal adenocarcinoma (PDAC), the lack of infiltrative ability of these CAR-T cells leads to sub-optimal treatment outcome. Chemokine (C-C motif) ligand 19 (CCL19)
Geng Liu et al.
Communications biology, 6(1), 102-102 (2023-01-27)
Protein acetylation plays a key role in regulating cellular processes and is subject to aberrant control in diverse pathologies. Although histone deacetylase (HDAC) inhibitors are approved drugs for certain cancers, it is not known whether they can be deployed in
ELISPOT Assay to Measure Peptide-specific IFN-γ Production.
Wykes M N and Renia L.
Bio-protocol, 7(11) (2017)
An optimized IFN-γ ELISpot assay for the sensitive and standardized monitoring of CMV protein-reactive effector cells of cell-mediated immunity.
Barabas S, et al.
BMC Immunology, 18(1), 14-14 (2017)
Daniel Larcombe-Young et al.
STAR protocols, 3(2), 101414-101414 (2022-05-28)
Dual co-stimulation may be harnessed using parallel chimeric antigen receptors (pCARs) in which two distinct co-stimulatory units are adjacently localized on the plasma membrane. This protocol summarizes construct design, human T cell isolation, retroviral transduction, tissue culture expansion, and preclinical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service