Skip to Content
Merck
All Photos(1)

Key Documents

47793

Supelco

trans-1,3-Dichloropropene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4Cl2
CAS Number:
Molecular Weight:
110.97
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

storage temp.

-10 to -25°C

SMILES string

[H]\C(Cl)=C(\[H])CCl

InChI

1S/C3H4Cl2/c4-2-1-3-5/h1-2H,3H2/b2-1+

InChI key

UOORRWUZONOOLO-OWOJBTEDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pablo I Nikel et al.
Philosophical transactions of the Royal Society of London. Series B, Biological sciences, 368(1616), 20120377-20120377 (2013-03-13)
Chlorinated pollutants are hardly biodegradable under oxic conditions, but they can often be metabolized by anaerobic bacteria through organohalide respiration reactions. In an attempt to identify bottlenecks limiting aerobic catabolism of 1,3-dichloroprop-1-ene (1,3-DCP; a widely used organohalide) in Pseudomonas pavonaceae
Pablo I Nikel et al.
Metabolic engineering, 15, 98-112 (2012-11-15)
Pseudomonas putida KT2440, a microbial cell factory of reference for industrial whole-cell biocatalysis, is unable to support biochemical reactions that occur under anoxic conditions, limiting its utility for a large number of relevant biotransformations. Unlike (facultative) anaerobes, P. putida resorts
Amit J Jhala et al.
Pest management science, 68(2), 225-230 (2011-07-29)
Preplant fumigation with methyl bromide (MeBr) has been used for control of soilborne pests in high-value annual, perennial and nursery crops, but is being phased out. In 2007 and 2008, research trials were conducted to evaluate the effects of surface
J E Thomas et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 46(8), 655-661 (2011-08-03)
Soil fumigation to control pests and pathogens is an important part of current agricultural practice. A reduction in fumigant emissions is required to ensure worker safety and environment health. A field trial in Florida was conducted to investigate whether carbonating
Qiuxia Wang et al.
Journal of agricultural and food chemistry, 58(1), 361-365 (2009-11-17)
The gelatin capsule (gel cap) formulation of 1,3-dichloropropene (1,3-D) was developed as a potential means of reducing 1,3-D emissions. The objective of this study was to determine the distribution, emission, and leaching of 1,3-D after applying the gel cap in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service