Skip to Content
Merck
All Photos(1)

Key Documents

47932

Supelco

EPA 502/524.2 VOC Mix

200 μg/mL each component in methanol, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Agency

EPA 502.2,524.1,524.2

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

200 μg/mL each component in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

General description

EPA 502/524.2 VOC Mix can be used for identifying and quantifying volatile halogenated organics and other volatile organics in drinking water. The analyst may be better equipped to choose between gas chromatography (GC) with a selective detector or gas chromatography/mass spectrometry (GC/MS) or high performance liquid chromatography (HPLC) as a preferred method of quantification. For US only, for EU: Cat. No. 32813.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Benzene
    Bromobenzene
    Bromochloromethane
    Bromodichloromethane
    Bromoform
    Bromomethane
    Butylbenzene
    sec-Butylbenzene
    tert-Butylbenzene
    Carbon tetrachloride
    Chloroethane
    Chloroform
    Chloromethane
    2-Chlorotoluene
    4-Chlorotoluene
    Dibromochloromethane
    1,2-Dibromo-3-chloropropane
    1,2-Dibromoethane
    Dibromomethane
    1,2-Dichlorobenzene
    1,3-Dichlorobenzene
    1,4-Dichlorobenzene
    Dichlorodifluoromethane
    1,1-Dichloroethane
    1,2-Dichloroethane
    1,1-Dichloroethylene
    cis-1,2-Dichloroethylene
    trans-1,2-Dichloroethylene
    Dichloromethane
    1,2-Dichloropropane
    1,3-Dichloropropane
    2,2-Dichloropropane
    1,1-Dichloro-1-propene
    cis-1,3-Dichloropropene
    trans-1,3-Dichloropropene
    Ethylbenzene
    Hexachloro-1,3-butadiene
    Naphthalene
    Propylbenzene
    Styrene
    1,1,1,2-Tetrachloroethane
    1,1,2,2-Tetrachloroethane
    Tetrachloroethylene
    Toluene
    1,2,3-Trichlorobenzene
    1,1,1-Trichloroethane
    1,1,2-Trichloroethane
    Trichloroethylene
    Trichlorofluoromethane
    1,2,3-Trichloropropane
    1,2,4-Trimethylbenzene
    Vinyl chloride
    1,2,4-Trichlorobenzene
    Cumene
    1,3,5-Trimethylbenzene solution
    o-Xylene
    m-Xylene
    p-Cymene
    p-Xylene
    See All (59)

Signal Word

Danger

Flash Point(F)

closed cup

Flash Point(C)

closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service