Skip to Content
Merck
All Photos(1)

Key Documents

47122

Supelco

Soybean oil

analytical standard

Synonym(s):

Soya oil from Glycine max

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12164500

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.4743 (lit.)

density

0.917 g/mL at 25 °C (lit.)

application(s)

food and beverages

format

neat

functional group

carboxylic acid
oleic acid

shipped in

ambient

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

Characterized soybean oil samples are available for use as controls or check samples for fatty acid methyl ester (FAME) analyses. Use the chromatographic fingerprint of our reference material to check for the presence of soybean oil in your sample. A Certificate of Composition which includes a chromatogram is shipped with each product.
These reference materials can be used as qualitative standards. They provide an excellent means of standardizing the lipid procedures and comparing the results to others. Packed in an amber ampule under nitrogen.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Soybean oil may be used as an analytical reference standard for the determination of the analyte in milk samples by high performance liquid chromatography technique.

Storage Class Code

10 - Combustible liquids

WGK

awg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Analysis of human milk triacylglycerols by high-performance liquid chromatography with light-scattering detection.
Pons SM, et al.
Journal of Chromatography A (1998)
Analysis of human milk triacylglycerols by high-performance liquid chromatography with light-scattering detection.
Pons SM, et al.
Journal of Chromatography A (1998)
Meera Penumetcha et al.
Lipids in health and disease, 12, 26-26 (2013-03-21)
Our previous work showed that dietary oxidized linoleic acid given, as a single fatty acid, to LDL receptor knockout mice decreased weight gain as compared to control mice. Other studies have also reported that animals fed oils heated for 24
The intralipid genie is out of the bottle-spin and wishful thinking.
N A Buckley et al.
Anaesthesia and intensive care, 41(2), 154-156 (2013-04-13)
Varadharajan Kavitha et al.
Indian journal of experimental biology, 51(5), 400-405 (2013-07-05)
The present study emphasizes in situ transformation of soybean oil to self-assembled supramolecular multilayered biopolymer material. The said polymer material was characterized and the entrapment efficacy of both hydrophilic and hydrophobic moieties was studied. In brief, soybean oil at varying

Articles

Learn more about Biodiesel as a renewable, alternative diesel fuel produced from vegetable oils, animal fats, or recycled restaurant grease.

Learn more about Biodiesel as a renewable, alternative diesel fuel produced from vegetable oils, animal fats, or recycled restaurant grease.

Learn more about Biodiesel as a renewable, alternative diesel fuel produced from vegetable oils, animal fats, or recycled restaurant grease.

Learn more about Biodiesel as a renewable, alternative diesel fuel produced from vegetable oils, animal fats, or recycled restaurant grease.

Chromatograms

application for HPLCapplication for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service