Skip to Content
Merck
All Photos(1)

Key Documents

S7381

Sigma-Aldrich

Soybean oil

dietary source of long-chain triglycerides and other lipids

Synonym(s):

Soya oil from Glycine max

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

biological source

soybean

form

liquid

refractive index

n20/D 1.4743 (lit.)

density

0.917 g/mL at 25 °C (lit.)
0.92 g/mL at 25 °C (lit.)

functional group

carboxylic acid
oleic acid

lipid type

oils

shipped in

ambient

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Soybean oil (SBO) is a vegetable oil widely used as biodegradable lubricant. It increases left ventricular contractility associated myosin ATPase and sarcoplasmic reticulum calcium pump (SERCA2a) in isolated hearts from rats. SBO is shown to reduce both total and non-HDL serum cholesterol and triglycerides induced by atherogenic diet in rats. It may be transesterified to biodiesel using CaO as a solid base catalyst. It may also be transesterified to biodiesel using methanol and ethanol.

Application

Soybean oil from Sigma is suitable for use:
  • in the determination of freezing point of soybean oil
  • as a standard at 0.1% in chloroform during the quantification of lipid by a vanillin-phosphoric acid reaction
  • as a delivery vehicle of α-lipoic acid
  • to dissolve tamoxifen
It may be used as a crude source for triglycerides and other lipids. Soybean oil is a potential raw material in oxygenated fatty acid production. It may be used in the synthesis of epoxidized-soybean oil (ESO).

Storage Class Code

10 - Combustible liquids

WGK

awg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Paulo J Oliveira et al.
Mitochondrion, 3(6), 337-345 (2005-08-27)
The heart is one of the organs affected during the later stages of diabetes. Mitochondrial function has already been proposed to be affected during the course of diabetes. Nevertheless, little information is known concerning the impact of antioxidants in heart
Christian Kaufmann et al.
Journal of insect physiology, 54(2), 367-377 (2007-12-08)
The role of adipokinetic hormones (AKHs) in the regulation of carbohydrate and lipid metabolism and flight performance was evaluated for females of the African malaria mosquito, Anopheles gambiae. Injection of various dosages of synthetic Anoga-AKH-I increased carbohydrate levels in the
Jonas Lexow et al.
Disease models & mechanisms, 6(6), 1470-1476 (2013-08-10)
Tamoxifen-inducible Cre-mediated manipulation of animal genomes has achieved wide acceptance over the last decade, with numerous important studies heavily relying on this technique. Recently, a number of groups have reported transient complications of using this protocol in the heart. In
Transesterification of soybean oil to biodiesel using CaO as a solid base catalyst.
Xuejun L, et al.
Fuel: The Science and Technology of Fuel and Energy, 87, 216-221 (2008)
A study of the oxidation and wear properties of vegetable oils: soybean oil without additives.
Castro W, et al.
Journal of the American Oil Chemists' Society, 83(1), 47-52 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service