Skip to Content
Merck
All Photos(1)

Key Documents

40061

Supelco

N-Nitrosodi-n-propylamine solution

certified reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116105

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

N(N=O)(CCC)CCC

InChI

1S/C6H14N2O/c1-3-5-8(7-9)6-4-2/h3-6H2,1-2H3

InChI key

YLKFDHTUAUWZPQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Moore et al.
Journal of the National Cancer Institute, 78(2), 289-293 (1987-02-01)
The effects of butylated hydroxyanisole [(BHA) CAS: 25013-16-5], alpha-tocopherol [(TC) CAS: 59-02-9], and carbazole [(CA) CAS: 86-74-8] administered subsequent to a single dose of 2,2'-dioxo-N-nitrosodipropylamine [(DOPN) CAS: 60599-38-4] on the development of putative preneoplastic lesions were investigated in Syrian golden
T Suzuki et al.
Mutation research, 444(2), 259-268 (1999-10-16)
We studied five carcinogens for (a) organ-specific mutagenicity and expression time in the transgenic (TG) mouse mutation assay and (b) clastogenicity in the peripheral blood micronucleus assay in the same mice. Groups of mice were injected intraperitoneally (ip) with N-nitroso-di-n-propylamine
L A Gritsiute et al.
Eksperimental'naia onkologiia, 11(3), 10-11 (1989-01-01)
187 female and male C57B1 mice received N-nitrosodipropylamine (NDPA) twice a week for 50 weeks by intragastric intubation. The total dose of NDPA administered to groups I and II was 3 mg or 1 mg per mouse. The main kinds
S Itoh et al.
Mutation research, 444(2), 309-319 (1999-10-16)
The mutagenic and clastogenic effects of N-nitrosodi-n-propylamine (NDPA) in lacZ transgenic mice (MutaMouse) were investigated as a part of the second collaborative study of the transgenic mouse mutation assay by a subgroup of the Mammalian Mutagenesis Study Group, a suborganization
N-Nitrosodi-n-propylamine.
Report on carcinogens : carcinogen profiles, 11, III200-III201 (2004-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service