Skip to Content
Merck
All Photos(1)

Key Documents

28564-U

Supelco

SLB®-5ms Capillary GC Column

L × I.D. 20 m × 0.18 mm, df 0.18 μm

Synonym(s):

GC column, SPB-5, 5% diphenyl, mass spec

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
eCl@ss:
32119290
NACRES:
SB.54
Pricing and availability is not currently available.

material

fused silica

Agency

Standard Method 6040D
EN 2005/108/EC
EPA 610,625,8015,8082,8100,8141,8270,OLM04.2 SVOA
EPA TO-13,IP-8,8270,525.2,608.1/608.2,608/8081/OLM04.2 PEST
JMHLW
NIOSH 2530,5503
OSHA 62
meets requirements for USP G27 and G36
suitable for EPA 1613

reg. compliance

FDA LIB 4423

parameter

-60-340 °C temperature (isothermal)
-60-360 °C temperature (programmed)

Beta value

250

df

0.18 μm

technique(s)

GC/MS: suitable
gas chromatography (GC): suitable (fast GC)

L × I.D.

20 m × 0.18 mm

matrix active group

Bonded and highly crosslinked; silphenylene polymer virtually equivalent in polarity to poly(5% diphenyl/95% dimethyl siloxane) phase

application(s)

agriculture
chemicals and industrial polymers
cleaning products
clinical
cosmetics
environmental
flavors and fragrances
food and beverages
forensics and toxicology
industrial hygiene
life science and biopharma
personal care
petroleum
pharmaceutical (small molecule)

column type

capillary non-polar

Looking for similar products? Visit Product Comparison Guide

General description

Application: The 5% phenyl equivalent phase provides a boiling point elution order with a slight increase in selectivity, especially for aromatic compounds. The low bleed characteristics, inertness, and durable nature make it the column of choice for environmental analytes (such as semivolatiles, pesticides, PCBs, and herbicides) or anywhere a low bleed non-polar column is required.
USP Code: This column meets USP G27 and G36 requirements.
Phase:
  • Bonded and highly crosslinked
  • Silphenylene polymer virtually equivalent in polarity to poly(5% diphenyl/95% dimethyl siloxane)
Temp. Limits:
  • ≤0.32 mm I.D.: -60 °C to 340 °C (isothermal) or 360 °C (programmed)
  • ≥0.53 mm I.D.: -60 °C to 330 °C (isothermal) or 340 °C (programmed)

Other Notes

We offer a variety of chromatography accessories including analytical syringes

Legal Information

SLB is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sophie Duclos et al.
Proteomics, 11(5), 854-864 (2011-02-01)
The essential roles of the endovacuolar system in health and disease call for the development of new tools allowing a better understanding of the complex molecular machinery involved in endocytic processes. We took advantage of the floating properties of small
Rasha M Faty et al.
Molecules (Basel, Switzerland), 20(2), 1842-1859 (2015-01-28)
Bromination of N-substituted homophthalimides and tetrahydropyrido[4,3-d]- pyrimidine-5,7-diones produces 4,4-dibromohomophthalimide and 8,8-dibromo-tetrahydropyrido[4,3-d]pyrimidine-5,7-dione derivatives, respectively, that can be used as precursors for spiro derivatives. The dibromo derivatives react with different binucleophilic reagents to produce several spiroisoquinoline and spirotetrahydropyrido[4,3-d]- pyrimidine-5,7-dione derivatives, respectively. Reaction
Annie Xu Zeng et al.
Analytica chimica acta, 803, 166-173 (2013-11-13)
Due to their distinct chemical properties, the application of ionic liquid (IL) compounds as gas chromatography (GC) stationary phases offer unique GC separation especially in the analysis of geometric and positional fatty acid methyl ester (FAME) isomers. Elution behaviour of
Lisa Simonsson et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 11(5), 1011-1017 (2010-03-20)
We investigate the Ca(2+)-triggered fusion of lipid vesicles site-selectively tethered to a DNA-modified supported lipid bilayer array, with the DNA strands designed such that hybridization occurs in a zipperlike fashion. Prior to the addition of Ca(2+), which is observed to
Frédéric Destaillats et al.
Journal of chromatography. A, 1218(52), 9384-9389 (2011-11-29)
Lipids found in human sebum contain specific fatty acids such as sapienic (cis-6 16:1), cis-8 18:1 and sebaleic (cis-5, cis-8 18:2) acids. These fatty acids belong to the n-10 series and the initial step involved in their synthesis is the

Articles

The Derivatization and Analysis of Amino Acids by GC-MS

The Derivatization and Analysis of Amino Acids by GC-MS

The Derivatization and Analysis of Amino Acids by GC-MS

The Derivatization and Analysis of Amino Acids by GC-MS

See All

Protocols

There are three types of analyses: GRO, DRO, and TPH. The published methods are simply variations of these to meet the different analytical needs of the individual agencies.

There are three types of analyses: GRO, DRO, and TPH. The published methods are simply variations of these to meet the different analytical needs of the individual agencies.

Chromatograms

suitable for GC, application for SPMEsuitable for GCsuitable for GCsuitable for GC, application for SPEShow More

Questions

1–6 of 6 Questions  
  1. Would you recommend SLB™-5ms Capillary GC Columns for all applications instead of Equity-5, SPB-5, PTE-5, MDN-5, and MDN-5S?

    1 answer
    1. Yes.

      Helpful?

  2. What if my method does not allow me to change to SLB™-5ms Capillary GC Columns?

    1 answer
    1. These methods typically specify a 5-type column (DB-5 or equivalent). The SLB-5ms satisfies that requirement.

      Helpful?

  3. Who uses SLB™-5ms Capillary GC Columns?

    1 answer
    1. Any analyst with the need for a non-polar column. Some applications include boiling point separations, GC-MS analyses of semivolatiles, and GC-ECD analyses of pesticides/PCBs. It is estimated that non-polar columns account for 51% of the worldwide column market.

      Helpful?

  4. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  5. How does the SLB™-5ms Capillary GC Column differ from our other 5-type columns?

    1 answer
    1. Many of our other 5-type columns (Equity-5, SPB-5, PTE-5, MDN-5, and MDN-5S) incorporate a siloxane polymer (backbone comprised of Si and O atoms only, side groups are either methyl or phenyl). The SLB-5ms column incorporates a silphenylene polymer (backbone comprised of Si and O atoms plus some phenyl groups, side groups are either methyl or phenyl).

      Helpful?

  6. What are the benefits of the SLB™-5ms Capillary GC Column over other manufacturers’ -5ms columns?

    1 answer
    1. The SLB-5ms excels in several performance criteria.· Low Bleed. The highly crosslinked silphenylene polymer chemistry reduces column bleed to a very low level, resulting in greater signal-to-noise level, allowing lower detection levels.· Inertness. The proprietary surface deactivation reduces interactions between analytes and the column surface, resulting in sharper peak shapes, allowing lower detection levels.· Durability. With the highest published maximum allowable operating temperature (MAOT) of any -5ms column, SLB-5ms columns are designed for long life.· Consistency. Innovative manufacturing processes result in decreased column variability, ensuring that the next column is as good as the last.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service