Skip to Content
Merck
All Photos(1)

Key Documents

21103AST

Supelco

Astec® CYCLOBOND I 2000 RSP Chiral (5 μm) HPLC Columns

L × I.D. 2 cm × 4 mm, HPLC Guard Cartridge

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

Astec® CYCLOBOND I 2000 RSP Chiral HPLC Guard, Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm

material

stainless steel column

Agency

suitable for USP L45

description

HPLC guard column

product line

Astec®

packaging

pkg of 1 ea

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

2 cm × 4 mm

matrix

silica particle platform
fully porous particle

matrix active group

cyclodextrin, beta- phase

particle size

5 μm

pore size

100 Å pore size

operating pH

3.5-7

separation technique

chiral

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chuong Pham-Huy et al.
Journal of biochemical and biophysical methods, 54(1-3), 287-299 (2003-01-25)
The enantioselective high-performance liquid chromatography (HPLC) of three racemic 3-hydroxybenzodiazepines, oxazepam (Oxa), lorazepam (Lor), and temazepam (Tem), is a difficult operation because of the spontaneous chiral inversion in polar solvent. To solve this problem, we have developed an HPLC method
Nilusha L T Padivitage et al.
Drug testing and analysis, 6(6), 542-551 (2013-10-12)
Recently a novel class of chiral stationary phases (CSPs) based on cyclofructan (CF) has been developed. Cyclofructans are cyclic oligosaccharides that possess a crown ether core and pendent fructofuranose moieties. Herein, we evaluate the applicability of these novel CSPs for
Jennifer L Donovan et al.
Free radical research, 40(10), 1029-1034 (2006-10-04)
Catechin is a flavonoid present in fruits, wine and cocoa products. Most foods contain the (+)-enantiomer of catechin but chocolate mainly contains ( - )-catechin, in addition to its major flavanol, ( - )-epicatechin. Previous studies have shown poor bioavailability
S Rudaz et al.
Chirality, 11(4), 319-325 (1999-05-29)
An extensive study of the behavior of three chiral stationary phases (CSP) is presented for the stereoselective determination of methadone. The following chromatographic columns were selected: a cellulose, Chiralcel OJ; a modified cyclodextrin. Cyclobond I 2000 RSP, and a protein
Hua He et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 814(2), 385-391 (2005-01-11)
The aim of this work was to present the advantages of two polymeric cartridges (Oasis HLB from Waters and Abselut Nexus from Varian) for the solid-phase extraction of methadone enantiomers and its major metabolite, 2-ethylidene-1,5-dimethyl-3,3-diphenylpyrrolidine (EDDP) and of some benzodiazepines

Related Content

CYCLOBOND is the name given to Astec technology for bonding cyclodextrins to a high purity silica gel through a stable ether linkage.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service