Skip to Content
Merck
All Photos(1)

Key Documents

20029-U

Supelco

ORBO 65P (OVS) Amberlite XAD®-4 specially cleaned, 160/80 mg

RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS), pkg of 10 ea

Synonym(s):

ORBO Amberlite XAD-4 Tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12190000
NACRES:
NB.24

material

RC,GFF,F,F (specially cleaned) separators

product line

Amberlite
ORBO

composition

Bed A, 160 mg
Bed B, 80 mg

packaging

pkg of 10 ea

manufacturer/tradename

ORBO 65P

technique(s)

active air sampling: suitable

O.D. × L

13-8 mm × 75 mm , OSHA Versatile Sampler (OVS)

matrix

XAD-4 (specially cleaned)

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for sampling Polar and non-polar VOCs; select aldehydes

Looking for similar products? Visit Product Comparison Guide

General description

ORBO 65P OSHA Versatile Sampler (OVS) is used for sampling both polar and non-polar volatile organic compounds (VOCs). It is particularly suitable for selecting aldehydes. The product is capped at the inlet and outlet to minimize environmental exposure.
It is commonly used as an adsorbent to collect and retain the volatile organic compound in the air samples, allowing for accurate determination of air concentrations.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.
ORBO is a trademark of Sigma-Aldrich Co. LLC
XAD is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Perfluorooctanesulfonate (PFOS) Conversion from N-Ethyl-N-(2-hydroxyethyl)-perfluorooctanesulfonamide (EtFOSE) in male Sprague Dawley rats after inhalation exposure.
Sue C, et al.
Environmental Research, 155, 307-313 (2017)
Bruno Laeng et al.
Cognitive processing, 12(1), 13-21 (2010-09-25)
We recorded the pupil diameters of participants performing the words' color-naming Stroop task (i.e., naming the color of a word that names a color). Non-color words were used as baseline to firmly establish the effects of semantic relatedness induced by
[Explorative abrasion--obsolete?].
A Orbo
Tidsskrift for den Norske laegeforening : tidsskrift for praktisk medicin, ny raekke, 111(19), 2397-2398 (1991-08-20)
K Flo et al.
Scandinavian journal of clinical and laboratory investigation, 55(8), 715-721 (1995-12-01)
Elevated extracellular cGMP levels have been observed in various clinical conditions, and the analyte has been proposed as a diagnostic marker of cardiovascular as well as malignant diseases. However, the use of extracellular cGMP as a pathophysiological marker requires detailed
Bjørn T Moe et al.
Anticancer research, 29(4), 1047-1052 (2009-05-06)
Endometrial hyperplasia is a precursor lesion of endometrial carcinoma. Clinical studies of endometrial hyperplasia have shown that levonorgestrel (LNG) is more therapeutically effective than medroxyprogesterone acetate (MPA). The present pharmacological in vitro study was performed to compare progestin effects on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service