Skip to Content
Merck
All Photos(1)

Key Documents

XAD4

Supelco

Amberlite XAD4 polymeric adsorbent

20-60 mesh

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000
NACRES:
SB.52
Pricing and availability is not currently available.

Product Name

Amberlite XAD4, 20-60 mesh

form

beads

Quality Level

autoignition temp.

800 °F

technique(s)

LPLC: suitable

surface area

750 m2/g

matrix

styrene-divinylbenzene

matrix active group

polymer

particle size

20-60 mesh

pore size

~0.98 mL/g pore volume
100 Å mean pore size

density

1.02 g/mL (true wet)(lit.)
1.08 g/mL (skeletal)(lit.)

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Nonionic macroreticular resin that adsorbs and releases ionic species through hydrophobic and polar interactions; usually used under isocratic conditions.

Application

Polyaromatic adsorbent for small hydrophobic compounds, surfactants, pharmaceutical manufacturing, phenols, chlorinated organics, pesticide removal and recovery, and organic removal from aqueous food streams.

Other Notes

Hydrophobic polyaromatic resin
Two particle sizes are available:
Mesh 20-60 are Cat. Nos. 20276, 10358, XAD4
Mesh 20-50 is Cat. No. 06444

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joanne Sh Tan et al.
Applied microbiology and biotechnology, 101(13), 5427-5437 (2017-05-10)
Ginsenosides are believed to be the principal components behind the pharmacological actions of ginseng, and their bioactive properties are closely related to the type, position, and number of sugar moieties attached to the aglycone; thus, modification of the sugar chains
Osman Çaylak et al.
Food chemistry, 274, 487-493 (2018-10-31)
An aminated Amberlite XAD-resin as a solid phase extractant was tested for speciation of inorganic mercury, Hg(II) and methylmercury, MeHg(I) in water and fish tissue samples. It was found that Hg(II) and MeHg(I) ions could be simultaneously retained by a
Marlen Kücklich et al.
PloS one, 12(8), e0183440-e0183440 (2017-08-26)
Previous studies showed that olfactory cues are important for mammalian communication. However, many specific compounds that convey information between conspecifics are still unknown. To understand mechanisms and functions of olfactory cues, olfactory signals such as volatile compounds emitted from individuals
ZhiRong Liu et al.
The journal of maternal-fetal & neonatal medicine : the official journal of the European Association of Perinatal Medicine, the Federation of Asia and Oceania Perinatal Societies, the International Society of Perinatal Obstetricians, 33(4), 693-705 (2018-10-26)
Introduction: We carried out a systematic review and meta-analysis to assess the safety and effectiveness of uterine artery embolization (UAE) compared with conventional hysterectomy on refractory postpartum hemorrhage (PPH).Methods: We searched PubMed, Embase, Chinese National Knowledge Infrastructure database (CNKI), Cochrane
Mariafrancesca Scalise et al.
International journal of molecular sciences, 19(3) (2018-03-03)
The human plasma membrane transporter ASCT2 is responsible for mediating Na- dependent antiport of neutral amino acids. New insights into structure/function relationships were unveiled by a combined approach of recombinant over-expression, site-directed mutagenesis, transport assays in proteoliposomes and bioinformatics. WT

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. What is the recommended method for cleaning XAD4 before use?

    1 answer
    1. The following pretreatment is recommended:
      1.The adsorbent is shipped as a water-wet product with sodium chloride (NaCl) and sodium carbonate (Na2CO3) salts to prevent bacterial growth. These salts need to be washed from the adsorbent before use, and it is suggested that this be achieved by washing with water at a linear flow rate of 5-10 m/h until the required level is achieved.
      2.For sensitive applications, residual monomeric or oligomeric compounds may need to be removed from the adsorbent.
      3.A regeneration with the proposed regenerant is also recommended before the first service cycle. If the regenerant is an alcohol, it must be displaced with water before the first loading cycle.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service