Z325295
BRAND® BLAUBRAND® bulb pipette, calibrated to deliver (TD, EX)
capacity 1 mL, two marks
About This Item
Recommended Products
material
AR-Glas®
description
partial delivery
feature
class AS
packaging
pack of 12 ea
manufacturer/tradename
BRAND 29722
volume measuring range
1 mL, accuracy: 0.008 mL
capacity
1 mL
Related Categories
General description
According to DIN EN ISO 648.
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Protocols
This page contains detailed information on working with and handling pipettes and pipetting aids.
This page contains detailed information on working with and handling pipettes and pipetting aids.
This page contains detailed information on working with and handling pipettes and pipetting aids.
This page contains detailed information on working with and handling pipettes and pipetting aids.