Key Documents
Z325007
BRAND® BLAUBRAND® bulb pipette, calibrated to deliver (TD, EX)
capacity 8 mL, one mark
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
material
AR-Glas®
description
total delivery Ex + 15 sec waiting time
feature
class AS
packaging
pack of 6 ea
manufacturer/tradename
BRAND 29710
capacity
8 mL
General description
Additional to DIN
All BLAUBRAND bulb pipettes are supplied with a batch number and an accompanying batch certificate in the original packaging.
According to DIN EN ISO 648.
Other Notes
BR29710 (India)
Legal Information
AR-Glas is a registered trademark of Schott AG
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Visit the Document Library
Protocols
This page contains detailed information on working with and handling pipettes and pipetting aids.