Skip to Content
Merck
All Photos(2)

Key Documents

Z1001

Sigma-Aldrich

Zinc sulfate heptahydrate

suitable for plant cell culture

Synonym(s):

Zinc Sulfate hydrate, ZnSO4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnSO4 · 7H2O
CAS Number:
Molecular Weight:
287.56
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.71
Assay:
>99.0%
solubility:
H2O: 100 mg/mL, clear, colorless

Assay

>99.0%

Quality Level

technique(s)

cell culture | plant: suitable

solubility

H2O: 100 mg/mL, clear, colorless

SMILES string

O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

InChI

1S/H2O4S.7H2O.Zn/c1-5(2,3)4;;;;;;;;/h(H2,1,2,3,4);7*1H2;/q;;;;;;;;+2/p-2

InChI key

RZLVQBNCHSJZPX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Use as source of Zn to support cell membrane stability and gene regulation.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zinc sulfate treatment of vocal process granuloma.
Taner Yilmaz
European archives of oto-rhino-laryngology : official journal of the European Federation of Oto-Rhino-Laryngological Societies (EUFOS) : affiliated with the German Society for Oto-Rhino-Laryngology - Head and Neck Surgery, 270(3), 1175-1175 (2012-10-12)
Acute diarrhea in adults and children: a global perspective.
Michael Farthing et al.
Journal of clinical gastroenterology, 47(1), 12-20 (2012-12-12)
Gaia Dell'Ariccia et al.
The Journal of experimental biology, 216(Pt 8), 1430-1433 (2013-01-12)
Olfactory cues have been shown to be important to homing petrels at night, but apparently those procellariiform species that also come back to the colony during the day are not impaired by smell deprivation. However, the nycthemeral distribution of homing
Dervis Özcelik et al.
Biological trace element research, 150(1-3), 342-349 (2012-10-12)
Zinc is an element that under physiological conditions preferentially binds to and is a potent inducer of metallothionein under physiological conditions. The present study was conducted to explore whether zinc supplementation morphologically and biochemically protects against diabetic nephropathy through modulation
Haichen Niu et al.
Pharmacology, biochemistry, and behavior, 103(3), 526-534 (2012-10-30)
Previous studies have shown that olfactory impairment by disrupting the olfactory epithelium prior to morphine administration attenuated the development addiction-related behaviors. However, it is unclear whether olfactory impairment will affect the expression of already established addiction-related behaviors. To address this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service