Skip to Content
Merck
All Photos(1)

Key Documents

X2254

Sigma-Aldrich

XE-991

≥98% (HPLC), powder, KCNQ channel blocker

Synonym(s):

10,10-bis(4-pyridinylmethyl)-9(10H)-anthracenone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H20N2O
CAS Number:
Molecular Weight:
376.45
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

XE-991, ≥98% (HPLC)

Assay

≥98% (HPLC)

color

white to beige

solubility

DMSO: >20 mg/mL

originator

Merck & Co., Inc., Kenilworth, NJ, U.S.

storage temp.

2-8°C

SMILES string

O=C1c2ccccc2C(Cc3ccncc3)(Cc4ccncc4)c5ccccc15

InChI

1S/C26H20N2O/c29-25-21-5-1-3-7-23(21)26(17-19-9-13-27-14-10-19,18-20-11-15-28-16-12-20)24-8-4-2-6-22(24)25/h1-16H,17-18H2

InChI key

KHJFBUUFMUBONL-UHFFFAOYSA-N

Related Categories

Application

XE-991 has been used as a KCNQ (Kv7.2/7.3) inhibitor to examine whether M-current inhibition affects oxytocin receptor (TGOT) mediated depolarization. It has also been used as an KCNQ inhibitor to study the ionic mechanism responsible for the overshoot/undershoot in membrane potential in mouse cholinergic interneurons (ChIs) in the presence of tetrodotoxin (TTX).

Biochem/physiol Actions

XE-991 is a KCNQ channel blocker.
XE-991 is a KCNQ channel blocker; which is more potent than linopiridine (Cat. No. L-134).

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Merck & Co., Inc., Kenilworth, NJ, U.S.. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lena Rubi et al.
Neuromolecular medicine, 15(3), 476-492 (2013-05-23)
Neuronal L-type voltage-gated calcium channels (LTCCs) are involved in several physiological functions, but increased activity of LTCCs has been linked to pathology. Due to the coupling of LTCC-mediated Ca(2+) influx to Ca(2+)-dependent conductances, such as KCa or non-specific cation channels
Pauses in cholinergic interneuron activity are driven by excitatory input and delayed rectification, with dopamine modulation
Zhang Y, et al.
Neuron, 98(5), 918-925 (2018)
David Bartolomé-Martín et al.
Neuropharmacology, 161, 107568-107568 (2019-03-20)
Dopamine and glutamate transporters (DAT and GLT-1, respectively) share some biophysical characteristics, as both are secondary active carriers coupled to electrochemical ion gradients. In order to identify common or specific components of their respective proteomes, we performed a proximity labelling
Oxytocin transforms firing mode of CA2 hippocampal neurons
Tirko N, et al.
Neuron, 100(3), 593-608 (2018)
Yusuf Olgar et al.
Journal of cellular physiology, 237(2), 1353-1371 (2021-10-12)
Insufficient-heart function is associated with myocardial insulin resistance in the elderly, particularly associated with long-QT, in a dependency on dysfunctional KCNQ1/KCNE1-channels. So, we aimed to examine the contribution of alterations in KCNQ1/KCNE1-current (IKs ) to the aging-related remodeling of the heart

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service